Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: CAIGVRCGRH ITSHGLALNC STDLTWFEHI VPCGLVGTGV TSLSKELQRH VTVEEVMPPF LVAFKEIYKC TLISEDS
Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 90%.
Catalyzes the transfer of endogenously produced octanoic acid from octanoyl-acyl-carrier-protein onto the lipoyl domains of lipoate-dependent enzymes. Lipoyl-ACP can also act as a substrate although octanoyl-ACP is likely to be the physiological substrate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Lipoate-protein ligase B; Lipoyl/octanoyl transferase; Lipoyltransferase 2; Octanoyl-[acyl-carrier-protein]-protein N-octanoyltransferase; Octanoyl-[acyl-carrier-protein]:protein N-octanoyltransferase LIPT2, mitochondrial; putative octanoyltransferase, mitochondrial
Gene Aliases: LIPT2
UniProt ID: (Human) A6NK58
Entrez Gene ID: (Human) 387787
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support