Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Positive test controls include: U-87MG, Mouse testis, Mouse brain.
Immunogen sequence: MVLSQRQRDE LNRAIADYLR SNGYEEAYSV FKKEAELDVN EELDKKYAGL LEKKWTSVIR LQKKVMELES KLNEAKEEFT SGGPLGQKRD PKEWIPRPPE
VAMP7 is a member of the soluble N-ethylmaleimide-sensitive factor attachment protein receptor (SNARE) family, localizing to late endosomes and lysosomes. VAMP7 is thought to mediate the fusion of endosomes to their target lysosomes as well as other exocytosis events during phagocytosis and neuritogenesis. VAMP7 interacts with the VPS9 ankyrin repeat protein VARP, a protein that localizes to early endosomes and thought to regulate endosome dynamics. Together with CD82, VAMP7 can modulate the signaling of EGFR by regulating its endocytosis from the plasma membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: LIS-1; LIS-1 {ECO:0000255|HAMAP-Rule:MF_03141}; lissencephaly 1 protein; Lissencephaly-1 protein; lissencephaly-1 protein {ECO:0000255|HAMAP-Rule:MF_03141}; MGC25297; PAF acetylhydrolase 45 kDa subunit; PAF acetylhydrolase 45 kDa subunit {ECO:0000255|HAMAP-Rule:MF_03141}; PAF-AH 45 kDa subunit; PAF-AH 45 kDa subunit {ECO:0000255|HAMAP-Rule:MF_03141}; PAF-AH alpha; PAF-AH alpha {ECO:0000255|HAMAP-Rule:MF_03141}; PAF-AH beta; PAFAH alpha; PAFAH alpha {ECO:0000255|HAMAP-Rule:MF_03141}; PAFAH1B1; platelet-activating factor acetylhydrolase 1b, regulatory subunit 1; platelet-activating factor acetylhydrolase 1b, regulatory subunit 1 (45kDa); platelet-activating factor acetylhydrolase beta subunit (PAF-AH beta); Platelet-activating factor acetylhydrolase IB subunit alpha; platelet-activating factor acetylhydrolase IB subunit alpha {ECO:0000255|HAMAP-Rule:MF_03141}; Platelet-activating factor acetylhydrolase IB subunit beta; platelet-activating factor acetylhydrolase, isoform 1b, beta1 subunit; platelet-activating factor acetylhydrolase, isoform 1b, subunit 1; platelet-activating factor acetylhydrolase, isoform Ib, alpha subunit (45kD); platelet-activating factor acetylhydrolase, isoform Ib, alpha subunit 45kDa; platelet-activating factor acetylhydrolase, isoform Ib, subunit 1 (45kDa); RP23-194P5.2
Gene Aliases: LIS-1; LIS1; LIS2; MDCR; MDS; Mdsh; MMS10-U; Ms10u; NudF; PAFAH; PAFAH1B1; PAFAHA
UniProt ID: (Human) P43034, (Rat) P63004, (Mouse) P63005
Entrez Gene ID: (Human) 5048, (Rat) 83572, (Mouse) 18472
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support