Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSVPGPYQA ATGPSSAPSA PPSYEETVAV NSYYPTPPAP MPGPTTGLVT GPDGKGMNPP SYYTQPAPIP NNNPITVQTV YVQHPITFLD RPIQMCCPSC NKMIVSQLSY NAGALTWLSC (1-119 aa encoded by BC008309)
Lipopolysaccharide is a potent stimulator of monocytes and macrophages, causing secretion of tumor necrosis factor-alpha (TNF-alpha) and other inflammatory mediators. This gene encodes lipopolysaccharide-induced TNF-alpha factor, which is a DNA-binding protein and can mediate the TNF-alpha expression by direct binding to the promoter region of the TNF-alpha gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CMT1C; FLJ38636; lipopolysaccharide-induced TNF factor; lipopolysaccharide-induced TNF-alpha factor; Lipopolysaccharide-induced tumor necrosis factor-alpha factor; LPS-induced TNF-alpha factor; MGC116698; MGC116700; MGC116701; MGC125274; MGC125275; MGC125276; p53-induced gene 7 protein; PIG 7; Small integral membrane protein of lysosome/late endosome; tumor protein p53 inducible protein 7
Gene Aliases: LITAF; PIG7; SIMPLE; TP53I7
UniProt ID: (Human) Q99732
Entrez Gene ID: (Human) 9516
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support