Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: GIASCVFTRH GQGFYLISPS EFERFSLSAR NITEPLCSLD INWPRDATQA SYRIRESPKL SQANGTPSIL LAPQSLDGSP DPAHSMGPDT PEPPEAALSP
LLGL1 is a protein that is similar to a tumor suppressor in Drosophila. The protein is part of a cytoskeletal network and is associated with nonmuscle myosin II heavy chain and a kinase that specifically phosphorylates this protein at serine residues. The gene for LLGL1 is located within the Smith-Magenis syndrome region on chromosome 17.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DLG4; Hugl protein; Hugl-1; Human homolog to the D-lgl gene protein; lethal giant larvae homolog 1; lethal giant larvae homolog 1, scribble cell polarity complex component; Lethal(2) giant larvae protein homolog 1; LLGL
Gene Aliases: DLG4; HUGL; HUGL-1; HUGL1; Lgl1; LLGL; LLGL1; Mgl1
UniProt ID: (Human) Q15334
Entrez Gene ID: (Human) 3996
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support