Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: SLKVKGGASE LQEDESFTLR GPPGAAPSAT QITVVLPHSS CELLYLGTES GNVFVVQLPA FRALEDRTIS SDAVLQRLPE EARHRRVFEM VEALQEHPR
LLGL1 is a protein that is similar to a tumor suppressor in Drosophila. The protein is part of a cytoskeletal network and is associated with nonmuscle myosin II heavy chain and a kinase that specifically phosphorylates this protein at serine residues. The gene for LLGL1 is located within the Smith-Magenis syndrome region on chromosome 17. LLGL2 is a protein similar to lethal (2) giant larvae of Drosophila. In fly, the protein's ability to localize cell fate determinants is regulated by the atypical protein kinase C (aPKC). In human, this protein interacts with aPKC-containing complexes and is cortically localized in mitotic cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HGL; human giant larvae homolog; lethal giant larvae homolog 2; lethal giant larvae homolog 2, scribble cell polarity complex component; Lethal(2) giant larvae protein homolog 2; LLGL scribble cell polarity complex component 2
Gene Aliases: HGL; Hugl-2; LGL2; LLGL2
UniProt ID: (Human) Q6P1M3
Entrez Gene ID: (Human) 3993
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support