Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MVTTLGPKMA AEWGGGVGYS GSGPGRSRWR WSGSVWVRSV LLLLGGLRAS ATSTPVSLGS SPPCRHHVPS DTEVINKVHL KANHVVKRDV DEHLRIKTVY DKSVEELLPE KKNLVKNKLF PQAISYLEKT FQVRRPAGTI LLSRQCATNQ YLRKENDPHR YCTGECAAHT KCGPVIVPEE HLQQCRVYRG GKWPHGAVGV PDQEGISDAD FVLYVGALAT ERCSHENIIS YAAYCQQEAN MDRPIAGYAN LCPNMISTQP QEFVGMLSTV KHEVIHALGF SAGLFAFYHD KDGNPLTSRF ADGLPPFNYS LGLYQWSDKV VRKVERLWDV RDNKIVRHTV YLLVTPRVVE EARKHFDCPV LEGMELENQG GVGTELNHWE KRLLENEAMT GSHTQNRVLS RITLALMEDT GRQMLSPYCD TLRSNPLQLT CRQDQRAVAV CNLQKFPKPL PQEYQYFDEL SGIPAEDLPY YGGSVEIADY CPFSQEFSWH LSGEYQRSSD CRILENQPEI FKNYGAEKYG PHSVCLIQKS AFVMEKCERK LSYPDWGSGC YQVSCSPQGL KVWVQDTSYL CSRAGQVLPV SIQMNGWIHD GNLLCPSCWD FCELCPPETD PPATNLTRAL PLDLCSCSSS LVVTLWLLLG NLFPLLAGFL LCIWH
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Invadolysin; leishmanolysin-2; leishmanolysin-like (metallopeptidase M8 family); Leishmanolysin-like peptidase
Gene Aliases: GP63; INV; IX14; LMLN; MSP
UniProt ID: (Human) Q96KR4
Entrez Gene ID: (Human) 89782
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support