Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VQPKGKQKGC AGCNRKIKDR YLLKALDKYW HEDCLKCACC DCRLGEVGST LYTKANLILC RRDYLRLFGT TGNCAACSKL IPAFEMVMRA RDNVYHLDCF ACQLCNQRFC VGDKFFLKNN MILCQMDYEE GQLNGTFESQ VQ
LMO1 encodes a cysteine-rich, two LIM domain transcriptional regulator. It is mapped to an area of consistent chromosomal translocation in chromosome 11, disrupting it in T-cell leukemia, although more rarely than the related gene, LMO2 is disrupted.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cysteine-rich protein TTG-1; LIM domain only 1 (rhombotin 1); LIM domain only protein 1; LMO-1; rhombotin 1; Rhombotin-1; T-cell translocation gene 1; T-cell translocation protein 1
Gene Aliases: LMO1; RBTN1; RHOM1; TTG1
UniProt ID: (Human) P25800
Entrez Gene ID: (Human) 4004
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support