Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: MRAKDNVYHL DCFACQLCNQ RFCVGDKFFL KNNMILCQTD YEEGLMKEGY APQVR*
The protein encoded by this gene belongs to the rhombotin family of cysteine-rich LIM domain oncogenes. This gene is predominantly expressed in the brain. Related family members, LMO1 and LMO2 on chromosome 11, have been reported to be involved in chromosomal translocations in T-cell leukemia. Many alternatively spliced transcript variants have been found for this gene. [provided by RefSeq, Aug 2011].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: LIM domain only 3 (rhombotin-like 2); LIM domain only protein 3; LMO-3; Neuronal-specific transcription factor DAT1; Rhombotin-3
Gene Aliases: LMO3; RBTN3; RBTNL2; Rhom-3; RHOM3
UniProt ID: (Human) Q8TAP4
Entrez Gene ID: (Human) 55885
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support