Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MLDGIKMEE HALRPGPATL GVLLGSDCPH PAVCEGCQRP ISDRFLMRVN ESSWHEECLQ CAACQQALTT SCYFRDRKLY CKQDYQQLFA AKCSGCMEKI APTEFVMRAL ECVYHLGCFC CCVCERQLRK GDEFVLKEGQ LLCKGDYEKE KDLLSSVSPD ESDSVKSEDE DGDMKPAKGQ GSQSKGSGDD GKDPRRPKRP RTILTTQQRR AFKASFEVSS KPCRKVRETL AAETGLSVRV VQVWFQNQRA KMKKLARRHQ QQQEQQNSQR LGQEVLSSRM EGMMASYTPL APPQQQIVAM EQSPYGSSDP FQQGLTPPQM PGNDSIFHDI DSDTSLTSLS DCFLGSSDVG SLQARVGNPI DRLYSMQSSY FAS (1-372 aa encoded by BC112120)
LMX1B, LIM homeobox transcription factor 1B, belongs to the LIM-homeodomain family. Members of this family are known to be important for pattern formation during development. LMX1B regulates mid-hindbrain patterning LMX1B-null mice embryos have a severe reduction in the number of midbrain dopaminergic neurons compared to wild-type. While LMX1B appears to be important for both the development and the survival of dopamine neurons, the related LMX1A is crucial for the differentiation of these cells. However, LMX1A and LMX1B function cooperatively to regulated the proliferation, specification and differentiation of midbrain dopaminergic neuronal progenitors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: LIM homeo box transcription factor 1B; LIM homeobox transcription factor 1 beta; LIM homeobox transcription factor 1, beta; LIM homeobox transcription factor 1-beta; LIM/homeobox protein 1.2; LIM/homeobox protein LMX1B; LMX-1.2
Gene Aliases: LMX1.1; LMX1.2; LMX1B; NPS1
UniProt ID: (Human) O60663, (Mouse) O88609
Entrez Gene ID: (Human) 4010, (Rat) 114501, (Mouse) 16917
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support