Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Control: Mouse brain tissue, rat spinal cord tissue.
LNP (LNPK) is an endoplasmic reticulum (ER)-shaping membrane protein that plays a role in determining ER morphology. It is involved in the stabilization of nascent three-way ER tubular junctions within the ER network and may play a role as a curvature-stabilizing protein within the three-way ER tubular junction network. LNP is involved in central nervous system development and may be involved in limb development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Endoplasmic reticulum junction formation protein lunapark; ER junction formation factor lunapark; lunapark; protein lunapark; Protein ulnaless
Gene Aliases: 2310011O18Rik; 4921514L11Rik; 9530051D01Rik; AI666268; Kiaa1715; Lnp; Lnpk; Ul; Uln; ulnaless
UniProt ID: (Mouse) Q7TQ95
Entrez Gene ID: (Mouse) 69605
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support