Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MASASQGADD DGSRRKPRLA ASLQISPQPR PWRPLPAQAQ SAWGPAPAPA TYRAEGGWPQ VSVLRDSGPG AGAGVGELGA ARAWENLGEQ MGKAPRVPVP PAGLSLPLKD PPASQAVSLL TEYAASLGIF LLFREDQPPG PCFPFSVSAE LDGVVCPAGT ANSKTEAKQQ AALSALCYIR SQLENPESPQ TSSRPPLAPL SVENILTHEQ RCAALVSAGF DLLLDERSPY WACKGTVAGV ILEREIPRAR GHVKEIYKLV ALGTGSSCCA GWLEFSGQQL HDCHGLVIAR RALLRFLFRQ LLLATQGGPK GKEQSVLAPQ PGPGPPFTLK PRVFLHLYIS NTPKGAARDI YLPPTSEGGL PHSPPMRLQA HVLGQLKPVC YVAPSLCDTH VGCLSASDKL ARWAVLGLGG ALLAHLVSPL YSTSLILADS CHDPPTLSRA IHTRPCLDSV LGPCLPPPYV RTALHLFAGP PVAPSEPTPD TCRGLSLNWS LGDPGIEVVD VATGRVKANA ALGPPSRLCK ASFLRAFHQA ARAVGKPYLL ALKTYEAAKA GPYQEARRQL SLLLDQQGLG AWPSKPLVGK FRN
ADAD2 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adenosine deaminase domain-containing protein 2; Testis nuclear RNA-binding protein-like
Gene Aliases: ADAD2; TENRL
UniProt ID: (Human) Q8NCV1
Entrez Gene ID: (Human) 161931
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support