Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: METGRQTGVS AEMLAMPRGL KGSKKDGIPE DLDGNLEAPR DQEGELRSED VMDLTEGDSE ASASAPPAAK RRKTHTKGKK ESKPTVDAEE AQRMTTLLSS MSEEQLSRYE VCRRSAFPRA RVAGLMRAIT GSSVSENAAI AMAGIAKLFV GEVVEEALDV CEMWGETPPL QPKHLREAVR RLKPKGLFPN SNCKRIMF
Predicted to enable transcription coactivator activity. Predicted to be involved in RNA polymerase II preinitiation complex assembly. Predicted to be part of transcription factor TFIID complex. [provided by Alliance of Genome Resources, Apr 2022]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: LOC391746
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support