Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPWLLSAPKL VPAVANVRGL SGCMLCSQRR YSLQPVPERR IPNRYLGQPS PFTHPHLLRP GEVTPGLSQV EYALRRHKLM SLIQKEAQGQ SGTDQTVVVL SNPTYYMSND IPYTFHQDNN FLYLCGFQEP DSILVLQSLP GKQLPSHKAI LFVPRRDPSR ELWDGPRSGT DGAIALTGVD EAYTLEEFQH LLPKMKAETN MVWYDWMRPS HAQLHSDYMQ PLTEAKAKSK NKVRGVQQLI QRLRLIKSPA EIERMQIAGK LTSQAFIETM FTSKAPVEEA FLYAKFEFEC RARGADILAY PPVVAGGNRS NTLHYVKNNQ LIKDGEMVLL DGGCESSCYV SDITRTWPVN GRFTAPQAEL YEAVLEIQRD CLALCFPGTS LENIYSMMLT LIGQKLKDLG IMKNIKENNA FKAARKYCPH HVGHYLGMDV HDTPDMPRSL PLQPGMVITI EPGIYIPEDD KDAPEKFRGL GVRIEDDVVV TQDSPLILSA DCPKEMNDIE QICSQAS
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aminopeptidase P3; APP3; Intermediate Cleaving Peptidase 55; X-Pro aminopeptidase 3; X-prolyl aminopeptidase (aminopeptidase P) 3, putative; X-prolyl aminopeptidase 3, mitochondrial; Xaa-Pro aminopeptidase 3
Gene Aliases: APP3; ICP55; NPHPL1; XPNPEP3
UniProt ID: (Human) Q9NQH7
Entrez Gene ID: (Human) 63929
Molecular Function:
metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support