Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGSEQSSEA ESRPNDLNSS VTPSPAKHRA KMDDIVVVAQ GSQASRNVSN DPDVIKLQEI PTFQPLLKGL LSGQTSPTNA KLEKLDSQQV LQLCLRYQDH LHQCAEAVAF DQNALVKRIK EMDLSVETLF SFMQERQKRY AKYAEQIQKV NEMSAILRRI QMGIDQTVPL LDRLNSMLPE GERLEPFSMK PDRELRL (1-196 aa encoded by BC013668)
LOH12CR1 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BLOC-1-related complex subunit 5; Loss of heterozygosity 12 chromosomal region 1; loss of heterozygosity, 12, chromosomal region 1; Myristoylated lysosomal protein; Myrlysin
Gene Aliases: BORCS5; LOH12CR1; LOH1CR12
UniProt ID: (Human) Q969J3
Entrez Gene ID: (Human) 118426
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support