Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
This antibody does not cross-react with recombinant human LOX‑1.
Reconstitute at 0.5 mg/mL in sterile PBS.
LOX-1 (lectin-like oxidized LDL receptor -1) is a 40-50 kDa type II membrane glycoprotein class E scavenger receptor expressed by endothelial cells, macrophages, activated platelets and smooth muscle cells. LOX-1 binds to a wide range of ligands, including oxidized low-density lipoprotein (oxLDL), hypochlorite modified high-density lipoprotein (HDL), aged /apoptotic cells, activated platelets and bacteria, reflecting its versatile physiological functions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hLOX 1; hLOX1; Lectin-like oxidized LDL receptor 1; lectin-like oxLDL receptor 1; Lectin-type oxidized LDL receptor 1; LOX 1; LOX-1; Ox-LDL receptor 1; oxidised low density lipoprotein (lectin-like) receptor 1; Oxidized low-density lipoprotein receptor 1; sLOX 1; soluble LOX 1
Gene Aliases: LOX-1; Lox1; Olr1; Scare1; SR-EI
UniProt ID: (Mouse) Q9EQ09
Entrez Gene ID: (Mouse) 108078
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support