Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Immunogen sequence: GHKASFCLED TSCDYGYHRR FACTAHTQGL SPGCYDTYGA DIDCQWIDIT DVKPGNYILK VSVNPSYLVP ESDYTNNVVR CDIRYTGHHA YASGCTISPY
Lysyl oxidase (LOX), a copper-containing amine oxidase, belongs to a heterogeneous family of enzymes that oxidize primary amine substrates to reactive aldehydes. It plays a vital role in the formation and repair of the extra cellular matrix. As well, LOX is a multifunctional enzyme having diverse biological functions such as developmental regulation, tumor suppression, cell motility and cellular senescence. The secreted form of LOX is responsible for the invasive properties of hypoxic human cancer cells. Thus, it is essential for hypoxia-induced metastasis and is a good therapeutic target for preventing and treating metastases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Lysyl oxidase; Lysyl oxidase (an H-rev gene with its expression down-regulated in HRAS-transformed rat 208F fibroblasts); MGC105112; Protein-lysine 6-oxidase; Ras excision protein; ras recision
Gene Aliases: AI893619; H-rev142; LOX; Rrg; Rrg1; TSC-160
UniProt ID: (Human) P28300, (Mouse) P28301
Entrez Gene ID: (Human) 4015, (Rat) 24914, (Mouse) 16948
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support