Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a member of the lysyl oxidase gene family. The prototypic member of the family is essential to the biogenesis of connective tissue, encoding an extracellular copper-dependent amine oxidase that catalyses the first step in the formation of crosslinks in collagens and elastin. A highly conserved amino acid sequence at the C-terminus end appears to be sufficient for amine oxidase activity, suggesting that each family member may retain this function. The N-terminus is poorly conserved and may impart additional roles in developmental regulation, senescence, tumor suppression, cell growth control, and chemotaxis to each member of the family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: LOL; Lysyl oxidase 2; Lysyl oxidase homolog 1; Lysyl oxidase-like protein 1
Gene Aliases: LOL; Lox2; LOXL; LOXL1
UniProt ID: (Human) Q08397, (Mouse) P97873
Entrez Gene ID: (Human) 4016, (Mouse) 16949, (Rat) 315714
Molecular Function:
oxidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support