Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The protein encoded by this gene is an endocytic receptor involved in several cellular processes, including intracellular signaling, lipid homeostasis, and clearance of apoptotic cells. In addition, the encoded protein is necessary for the A2M-mediated clearance of secreted amyloid precursor protein and beta-amyloid, the main component of amyloid plaques found in Alzheimer patients. Expression of this gene decreases with age and has been found to be lower than controls in brain tissue from Alzheimer patients.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A2MR; alpha 2-macroglobulin receptor; Alpha-2-macroglobulin receptor; CD91; FLJ16451; lipoprotein receptor-related protein; LRP-1; lrp1; MGC88725; Prolow-density lipoprotein receptor-related protein 1
Gene Aliases: A2mr; AI316852; b2b1554Clo; CD91; Lrp; Lrp1
UniProt ID: (Mouse) Q91ZX7
Entrez Gene ID: (Mouse) 16971
Molecular Function:
apolipoprotein
extracellular matrix structural protein
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support