Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes the major vault protein which is a lung resistance-related protein. Vaults are multi-subunit structures that may be involved in nucleo-cytoplasmic transport. This protein mediates drug resistance, perhaps via a transport process. It is widely distributed in normal tissues, and overexpressed in multidrug-resistant cancer cells. The protein overexpression is a potentially useful marker of clinical drug resistance. This gene produces two transcripts by using two alternative exon 2 sequences; however, the open reading frames are the same in both transcripts.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Lung resistance-related protein; Major vault protein; MVP; MVP1; Testicular Secretory Protein Li 30; VAULT1; vRNP
Gene Aliases: 2310009M24Rik; cb771; LRP; MVP; VAULT1; wu:fb52b05; wu:fc02g01
UniProt ID: (Human) Q14764, (Rat) Q62667, (Mouse) Q9EQK5, (Zebrafish) Q6P3L0
Entrez Gene ID: (Human) 9961, (Rat) 64681, (Mouse) 78388, (Zebrafish) 373081
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support