Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: CSHLCLAVPV GGFVCGCPAH YSLNADNRTC SAPTTFLLFS QKSAINRMVI DEQQSPDIIL PIHSLRNVRA IDYDPLDKQL YWIDSRQNMI RKAQEDGSQG F
LRP6 is essential for the Wnt/beta catenin signaling pathway, probably by acting as a coreceptor together with Frizzled for Wnt. It is a specific, high-affinity receptor for DKK1 and DKK2, but not DKK3. The interaction with DKK1 blocks LRP6-mediated Wnt/beta catenin signaling via LRP6 removal via Kremen proteins-mediated endocytosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: crooked; FLJ90062; FLJ90421; low density lipoprotein receptor-related protein 6; low density lipoprotein receptor-related protein 6; low density lipoprotein-related protein 6; low density lipoprotein-related protein 6; Low-density lipoprotein receptor-related protein 6; lpr6; LRP-6
Gene Aliases:
ADCAD2; C030016K15Rik; Cd; Gw; LRP6; ska26; ska
UniProt ID: (Human) O75581, (Mouse) O88572
Entrez Gene ID: (Human) 4040, (Rat) 312781, (Mouse) 16974
Molecular Function:
apolipoprotein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support