Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive Control: Hela cell lysates, human colon tissue.
Subcellular Location: Endoplasmic reticulum, Cell membrane, Membrane raft.
LRP6 is essential for the Wnt/beta catenin signaling pathway, probably by acting as a coreceptor together with Frizzled for Wnt. It is a specific, high-affinity receptor for DKK1 and DKK2, but not DKK3. The interaction with DKK1 blocks LRP6-mediated Wnt/beta catenin signaling via LRP6 removal via Kremen proteins-mediated endocytosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ90062; FLJ90421; low density lipoprotein receptor-related protein 6; Low-density lipoprotein receptor-related protein 6; lpr6; LRP-6
Gene Aliases: ADCAD2; LRP6; STHAG7
UniProt ID: (Human) O75581
Entrez Gene ID: (Human) 4040
Molecular Function:
apolipoprotein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support