Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: YSREKNQPK PSPKRESGEE FRMEKLNQLW EKAQRLHLPP VRLAELHADL KIQERDELAW KKLKLDGLDE DGEKEARLIR NLNVILAKYG LDGKKDARQV TSNSLSGTQE DGLDDPRLEK LWHKAKTSGK FSGEELDKLW REFLHHKEKV HEYNVLLETL SRTEEIHENV ISPSDLSDIK GSVLHSRHTE LKEKLRSINQ GLDRLRRVSH QGYSTEAEFE EPRVIDLWDL AQSANLTDKE LEAFREELKH FEAKIEKHNH YQKQLEIAHE KLRHAESVGD GERVSRSREK HALLEGRTKE LGYTVKKHLQ DLSGRISRAR HNEL (35-357 aa encoded by BC112067)
Interacts with LRP1/alpha-2-macroglobulin receptor and glycoprotein 330.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 39 kDa receptor-associated protein; Alpha-2-macroglobulin receptor-associated protein; Alpha-2-MRAP; low density lipoprotein receptor-related protein associated protein 1; Low density lipoprotein receptor-related protein-associated protein 1; low density lipoprotein-related protein-associated protein 1 (alpha-2-macroglobulin receptor-associated protein 1); RAP
Gene Aliases: A2MRAP; A2RAP; alpha-2-MRAP; HBP44; LRPAP1; MRAP; MYP23; RAP
UniProt ID: (Human) P30533
Entrez Gene ID: (Human) 4043
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support