Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
LRRC47 gene ontology annotations related to this gene include phenylalanine-tRNA ligase activity; RNA binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Leucine-rich repeat-containing protein 47; RP1-286D6.3
Gene Aliases: 2900010D03Rik; AA960559; KIAA1185; LRRC47; mKIAA1185
UniProt ID: (Human) Q8N1G4, (Mouse) Q505F5
Entrez Gene ID: (Human) 57470, (Mouse) 72946
Molecular Function:
aminoacyl-tRNA synthetase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support