Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The protein encoded by this gene is cilium-specific and is required for the stability of the ciliary architecture. It is involved in the regulation of microtubule-based cilia and actin-based brush border microvilli. Mutations in this gene are associated with primary ciliary dyskinesia-13.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dynein (axonemal) assembly factor 1; Dynein axonemal assembly factor 1; dynein, axonemal, assembly factor 1; leucine rich repeat containing 50; Leucine-rich repeat-containing protein 50; outer row dynein assembly 7 homolog; testicular tissue protein Li 110
Gene Aliases: CILD13; DNAAF1; LRRC50; ODA7
UniProt ID: (Human) Q8NEP3
Entrez Gene ID: (Dog) 479628, (Human) 123872
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support