Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEERERGAR SAGAGSPARP PSPRLDVSSD SFDPLLALYA PRLPPIPYPN APCFNNVAEY ESFLRTGVRG GGRGRGRARG AAAGSGVPAA PGPSGRTRRR PDAPAPDPER IQRLRRLMVA KEEGDGAAGA GRRGPGRSRK APRNVLTRMP LHEGSPLGEL HRCIREGVKV NVHIRTFKGL RGVCTGFLVA FDKFWNMALT DVDETYRKPV LGKAYERDSS LTLTRLFDRL KLQDSSKKEA DSKSAVEDST LSRYSQTSTW KLASVWGRAD TGRGSHKRSR SVPSSLQASA REESRSELSG RTTRTDGSSV GGTFSRATTL SRGQSRKKKR KPKVDYQQVF TRHINQIFIR GENVLLVHLA Q (1-360 aa encoded by BC126449)
LSm11 is a 360 amino acid protein that localizes to the nucleus and belongs to the LSm subfamily of snRNP Sm proteins. Containing an N-terminal domain that is necessary for pre-mRNA cleavage, LSm11 functions to binds specifically to U7 snRNA and is thought to play a role in cell cycle regulation and cell growth. The gene encoding LSm11 maps to human chromosome 5, which contains 181 million base pairs and comprises nearly 6% of the human genome. Deletion of the p arm of chromosome 5 leads to Cri du chat syndrome, while deletion of the q arm or of chromosome 5 altogether is common in therapy-related acute myelogenous leukemias and myelodysplastic syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ38273; U7 snRNA-associated Sm-like protein LSm11
Gene Aliases: LSM11
UniProt ID: (Human) P83369
Entrez Gene ID: (Human) 134353
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support