Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SSTSSFQSM GSYGPFGRMP TYSQFSPSSL VGQQFGAVGV AGSSLTSFGT ETSNSGTLPQ SSAVGSAFTQ DTRSLKTQLS QGRSSPQLDP LRKSPTMEQA VQTASAHLPA PAAVGRRSPV STRPLPSASQ KAGENQEHRR AEVHKVSRPE NEQLRNDNKR QVAPGAPSAP RRGRGGHRGG RGRFGIRRDG PMKFEKDFDF ESANAQFNKE EIDREFHNKL KLKEDKLEKQ EKPVNGEDKG DSGVDTQNSE GNADEEDPLG PNCYYDKTKS FFDNISCDDN RERRPTWAEE RRLNAETFGI PLRPNRGRGG YRGRGGLGFR GGRGRGGGRG GTFTAPRGFR GGFRGGRGGR EFADFEYRKD NKVAA (100-463 aa encoded by BC016842)
LSM14A, Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family. Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AlphaSNBP; family with sequence similarity 61, member A; hRAP55; hRAP55A; LSM14 homolog A; LSM14A mRNA processing body assembly factor; LSM14A, SCD6 homolog A; Protein FAM61A; Protein LSM14 homolog A; Protein SCD6 homolog; Putative alpha-synuclein-binding protein; RNA-associated protein 55; RNA-associated protein 55A
Gene Aliases: C19orf13; FAM61A; LSM14A; RAP55; RAP55A
UniProt ID: (Human) Q8ND56
Entrez Gene ID: (Human) 26065
Molecular Function:
RNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support