Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LVLPPSFPY GGMENPCLTF VTPTLLAGDK SLSNVIAHEI SHSWTGNLVT NKTWDHFWLN EGHTVYLERH ICGRLFGEKF RHFNALGGWG ELQNSVKTFG ETHPFTKLVV DLTDIDPDVA YSSVPYEKGF ALLFYLEQLL GGPEIFLGFL KAYVEKFSYK SITTDDWKDF LYSYFKDKVD VLNQVDWNAW LYSPGLPPIK PNYDMTLTNA CIALSQRWIT AKEDDLNSFN ATDLKDLSSH QLNEFLAQTL QRAPLPLGHI KRMQEVYNFN AINNSEIRFR WLRLCIQSKW EDAIPLALKM ATEQGRMKFT RPLFKDLAAF DKSHDQAVRT YQEHKASMHP VTAMLVGKDL KVD (260-611 aa encoded by BC032528)
Hydrolyzes an epoxide moiety of leukotriene A4 (LTA-4) to form leukotriene B4 (LTB-4). The enzyme also has some peptidase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Leukotriene A(4) hydrolase; Leukotriene A-4 hydrolase; LKHA4; LTA-4 hydrolase; LTA4 hydrodase; testicular secretory protein Li 27; Tripeptide aminopeptidase LTA4H
Gene Aliases: LTA4; LTA4H
UniProt ID: (Human) P09960, (Mouse) P24527, (Rat) P30349
Entrez Gene ID: (Human) 4048, (Mouse) 16993, (Rat) 299732
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support