Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MLLPWATSAP GLAWGPLVLG LFGLLAASQP QAVPPYASEN QTCRDQEKEY YEPQHRICCS RCPPGTYVSA KCSRIRDTVC ATCAENSYNE HWNYLTICQL CRPCDPVMGL EEIAPCTSKR KTQCRCQPGM FCAAWALECT HCELLSDCPP GTEAELKDEV GKGNNHCVPC KAGHFQNTSS PSARCQPHTR CENQGLVEAA PGTAQSDTTC KNPLEPLPPE MSGTMLMLAV LLPLAFFLLL ATVFSCIWKS HPSLCRKLGS LLKRRPQGEG PNPVAGSWEP PKAHPYFPDL VQPLLPISGD VSPVSTGLPA APVLEAGVPQ QQSPLDLTRE PQLEPGEQSQ VAHGTNGIHV TGGSMTITGN IYIYNGPVLG GPPGPGDLPA TPEPPYPIPE EGDPGPPGLS TPHQEDGKAW HLAETEHCGA TPSNRGPRNQ FITHD
The protein encoded by this gene is a member of the tumor necrosis factor (TNF) family of receptors. It is expressed on the surface of most cell types, including cells of epithelial and myeloid lineages, but not on T and B lymphocytes. The protein specifically binds the lymphotoxin membrane form (a complex of lymphotoxin-alpha and lymphtoxin-beta). The encoded protein and its ligand play a role in the development and organization of lymphoid tissue and tranformed cells. Activation of the encoded protein can trigger apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: lymphotoxin B receptor; lymphotoxin beta receptor (TNFR superfamily, member 3); Lymphotoxin-beta receptor; TNF-RIII; TNFR-III; Tumor necrosis factor C receptor; Tumor necrosis factor receptor 2-related protein; Tumor necrosis factor receptor superfamily member 3; Tumor necrosis factor receptor type III
Gene Aliases: CD18; D12S370; LT-BETA-R; LTBR; TNF-R-III; TNFCR; TNFR-RP; TNFR2-RP; TNFR3; TNFRSF3
UniProt ID: (Human) P36941
Entrez Gene ID: (Human) 4055
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support