Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KDITRKDAE RQLLAPGNSA GAFLIRESET LKGSFSLSVR DFDPVHGDVI KHYKIRSLDN GGYYISPRIT FPCISDMIKH YQKQADGLCR RLEKACISPK PQKPWDKDAW EIPRESIKLV KRLGAGQFGE VWMGYYNNST KVAVKTLKPG TMSVQAFLEE ANLMKTLQHD KLVRLYAVVT REEPIYIITE YMAKGSLLDF LKSDEGGKVL LPKLIDFSAQ IAEGMAYIER KNYIHRDLRA ANVLVSESLM CKIADFGLAR VIEDNEYTAR EGAKFPIKWT APEAINFGCF TIKSDVWSFG ILLYEIVTYG KIPYPGRTNA DVMTALSQGY RMPRVENCPD ELYDIMKMCW KEKAEERPTF DYLQSVLDDF YTATEGQYQQ QP (132-512 aa encoded by BC075002)
Lyn proto oncogene (Lcl/Yes related novel protein tyrosine kinase) is encoded by the Lyn gene located on chromosome 8 in humans and belongs to the Src family kinase. It is a non-receptor tyrosine kinase that acts as a mediator in transducing cellular signaling. The most well studied expression of Lyn is in hematopoietic cell types, both lymphoid and myeloid cells with exclusion in T lymphocytes. It has also been observed that Lyn plays diverse roles such as in regulation of B cell receptor signaling, mast cell signaling and Epithelial - Mesenchymal Transition (EMT).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ26625; Lck/Yes-related novel protein tyrosine kinase; p53Lyn; p56Lyn; Tyrosine-protein kinase Lyn; V-yes-1 Yamaguchi sarcoma viral related oncogene homolog
Gene Aliases: JTK8; LYN; p53Lyn; p56Lyn
UniProt ID: (Human) P07948
Entrez Gene ID: (Human) 4067
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support