Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MWVLGIAATF CGLFLLPGFA LQIQCYQCEE FQLNNDCSSP EFIVNCTVNV QDMCQKEVME QSAGIMYRKS CASSAACLIA SAGYQSFCSP GKLNSVCISC CNTPLCNGPR PKKRGSSASA LRPGLRTTIL FLKLALFSAH C
Believed to act as a modulator of nicotinic acetylcholine receptors (nAChRs) activity. In vitro increases receptor desensitization and decreases affinity for ACh of alpha-4:beta-2- containing nAChRs. May play a role in the intracellular trafficking of alpha-4:beta-2 and alpha-7-containing nAChRs and may inhibit their expression at the cell surface. May be involved in the control of anxiety.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ly-6/neurotoxin-like protein 2; Ly6/PLAUR domain-containing protein 1; Lynx2; PHTS; Putative HeLa tumor suppressor
Gene Aliases: 2700050C12Rik; AI853408; C530008O16Rik; Lynx2; LYPD1; LYPDC1; MNCb-0671; PHTS; PSEC0181; UNQ3079/PRO9917
UniProt ID: (Human) Q8N2G4, (Rat) Q66H42, (Mouse) Q8BLC3
Entrez Gene ID: (Human) 116372, (Rat) 360838, (Mouse) 72585
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support