Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Positive test controls include: Jurkat, HeLa, 293T, HepG2, Mouse brain, Mouse liver, Mouse pancreas. The target is usually found in the following locations: Cytoplasm, Endoplasmic reticulum membrane, Membrane, Nucleus, Single-pass type II membrane protein.
Immunogen sequence: MELELDAGDQ DLLAFLLEES GDLGTAPDEA VRAPLDWALP LSEVPSDWEV DDLLCSLLSP PASLNILSSS NPCLVHHDHT YSLPRETVSM DLESESCRKE GTQMTPQHME ELAEQEIARL VLTDEEKSLL EKEGLILPET LPLTKTEEQI LKRVRRKIRN KRSAQESRRK KKVYVGGLES RVLKYTAQNM ELQNKVQLLE EQNLSLLDQL RKLQAMVIEI SNKTSSSSTC
This gene encodes a transcription factor that is a member of the leucine zipper family of DNA binding proteins. This protein binds to the cAMP-responsive element, an octameric palindrome. The protein interacts with host cell factor C1, which also associates with the herpes simplex virus (HSV) protein VP16 that induces transcription of HSV immediate-early genes. This protein and VP16 both bind to the same site on host cell factor C1. It is thought that the interaction between this protein and host cell factor C1 plays a role in the establishment of latency during HSV infection. An additional transcript variant has been identified, but its biological validity has not been determined.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: basic leucine zipper protein; cAMP-responsive element-binding protein 3; CREB-3; cyclic AMP response element (CRE)-binding protein/activating transcription factor 1; Cyclic AMP-responsive element-binding protein 3; leucin zipper proitein; Leucine zipper protein; Luman; small leucine zipper protein; transcription factor LZIP; Transcription factor LZIP-alpha
Gene Aliases: AU044960; AW538053; C80076; CREB3; LUMAN; LZIP; LZIP-1; LZIP-2; sLZIP
UniProt ID: (Human) O43889
Entrez Gene ID: (Human) 10488, (Mouse) 12913, (Rat) 298400
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support