Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody detects endogenous levels of total LZK / MAP3K13 protein.
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The tyrosine-like kinase (TLK) group consists of 40 tyrosine and serine-threonine kinases such as MLK (mixed-lineage kinase), LISK (LIMK/TESK), IRAK (interleukin-1 receptor-associated kinase), Raf, RIPK (receptor-interacting protein kinase), and STRK (activin and TGF-beta receptors) families.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Leucine zipper-bearing kinase; MAP3K13; mitogen activated protein kinase kinase kinase 13; Mitogen-activated protein kinase kinase kinase 13; Mixed lineage kinase; MLK
Gene Aliases: C130026N12Rik; LZK; MAP3K13; MEKK13; MLK
UniProt ID: (Human) O43283, (Mouse) Q1HKZ5
Entrez Gene ID: (Human) 9175, (Rat) 303823, (Mouse) 71751
Molecular Function: non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support