Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The function remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: leucine zipper transcription factor-like 1; Leucine zipper transcription factor-like protein 1
Gene Aliases: 5530402H04Rik; 6130400H19Rik; AI414725; AW048545; BBS17; LZTFL1
UniProt ID: (Human) Q9NQ48, (Mouse) Q9JHQ5
Entrez Gene ID: (Human) 54585, (Mouse) 93730
Molecular Function:
basic leucine zipper transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support