Product References
Mongolian medicine Wenguanmu ointment treats eczema by inhibiting the CKLF-1/NF-κB pathway.
Journal of ethnopharmacology
Zhu L,Li XJ,Gu C,Gao Y,Zhang CS,Wang LY,Chen NH,Li G
Fri Sep 15 00:00:00 EDT 2023
ZSWIM1 Promotes the Proliferation and Metastasis of Lung Adenocarcinoma Cells through the STK38/MEKK2/ERK1/2 Axis.
Journal of proteome research
Gao X,Lian Q,Guan B,Liu QY,Meng M,Chen Y,Jin J,Li H,Liu X,Sun Z,Liu L,He QY,Zhang G
Fri Apr 07 00:00:00 EDT 2023
NamiRNA-enhancer network of miR-492 activates the NR2C1-TGF-β/Smad3 pathway to promote epithelial-mesenchymal transition of pancreatic cancer.
Carcinogenesis
Liu S,He X,Di Y,Li Q,Li F,Ma Y,Chen L,Gao Y,Xu J,Yang S,Xu L,Corpe C,Ling Y,Zhang X,Xu J,Yu W,Wang J
Fri May 26 00:00:00 EDT 2023
HnRNP K reduces viral gene expression by targeting cytosine-rich sequences in porcine reproductive and respiratory syndrome virus-2 genome to dampen the viral growth.
Virology
Jing H,Song Y,Li H,Duan E,Liu J,Ke W,Tao R,Li Y,Zhao P,Wang J,Cao S,Wang H,Sun Y,Zhang Y
Sat Apr 01 00:00:00 EDT 2023
LCK facilitates DNA damage repair by stabilizing RAD51 and BRCA1 in the nucleus of chemoresistant ovarian cancer.
Journal of ovarian research
Dey G,Bharti R,Braley C,Alluri R,Esakov E,Crean-Tate K,McCrae K,Joehlin-Price A,Rose PG,Lathia J,Gong Z,Reizes O
Tue Jun 27 00:00:00 EDT 2023
SDC1-dependent TGM2 determines radiosensitivity in glioblastoma by coordinating EPG5-mediated fusion of autophagosomes with lysosomes.
Autophagy
Zheng W,Chen Q,Liu H,Zeng L,Zhou Y,Liu X,Bai Y,Zhang J,Pan Y,Shao C
Wed Mar 01 00:00:00 EST 2023
Alternative transcribed 3' isoform of long non-coding RNA Malat1 inhibits mouse retinal oxidative stress.
iScience
Ghanam AR,Ke S,Wang S,Elgendy R,Xie C,Wang S,Zhang R,Wei M,Liu W,Cao J,Zhang Y,Zhang Z,Xue T,Zheng Y,Song X
Fri Jan 20 00:00:00 EST 2023
Alternative transcribed 3' isoform of long non-coding RNA Malat1 inhibits mouse retinal oxidative stress.
iScience
Ghanam AR,Ke S,Wang S,Elgendy R,Xie C,Wang S,Zhang R,Wei M,Liu W,Cao J,Zhang Y,Zhang Z,Xue T,Zheng Y,Song X
Fri Jan 20 00:00:00 EST 2023
LAP1 supports nuclear adaptability during constrained melanoma cell migration and invasion.
Nature cell biology
Jung-Garcia Y,Maiques O,Monger J,Rodriguez-Hernandez I,Fanshawe B,Domart MC,Renshaw MJ,Marti RM,Matias-Guiu X,Collinson LM,Sanz-Moreno V,Carlton JG
Sun Jan 01 00:00:00 EST 2023
LAP1 supports nuclear adaptability during constrained melanoma cell migration and invasion.
Nature cell biology
Jung-Garcia Y,Maiques O,Monger J,Rodriguez-Hernandez I,Fanshawe B,Domart MC,Renshaw MJ,Marti RM,Matias-Guiu X,Collinson LM,Sanz-Moreno V,Carlton JG
Sun Jan 01 00:00:00 EST 2023
Erxian decoction alleviates cisplatin-induced premature ovarian failure in rats by reducing oxidation levels in ovarian granulosa cells.
Journal of ethnopharmacology
Liu J,Yang Y,He Y,Feng C,Ou H,Yang J,Chen Y,You F,Shao B,Bao J,Guan X,Chen F,Zhao P
Sat Mar 25 00:00:00 EDT 2023
A small-molecule inhibitor of Keap1-Nrf2 interaction attenuates sepsis by selectively augmenting the antibacterial defence of macrophages at infection sites.
EBioMedicine
Wang Y,Tang B,Li H,Zheng J,Zhang C,Yang Z,Tan X,Luo P,Ma L,Wang Y,Long L,Chen Z,Xiao Z,Ma L,Zhou J,Wang Y,Shi C
Sat Apr 01 00:00:00 EDT 2023
Lycium barbarum polysaccharide alleviates dextran sodium sulfate-induced inflammatory bowel disease by regulating M1/M2 macrophage polarization via the STAT1 and STAT6 pathways.
Frontiers in pharmacology
Wang J,Gao H,Xie Y,Wang P,Li Y,Zhao J,Wang C,Ma X,Wang Y,Mao Q,Xia H
Tue Aug 08 00:00:00 EDT 2023
Proliferating cell nuclear antigen impairs the nuclear import of influenza A virus PB2 and suppresses virus replication.
Journal of medical virology
Zhou Y,Zhang S,Wang B,Ran Y,Tu S,Lin Z,Jin M,Chen H,Zou J,Zhou H
Thu Jun 01 00:00:00 EDT 2023
Progerinin, an Inhibitor of Progerin, Alleviates Cardiac Abnormalities in a Model Mouse of Hutchinson-Gilford Progeria Syndrome.
Cells
Kang SM,Seo S,Song EJ,Kweon O,Jo AH,Park S,Woo TG,Kim BH,Oh GT,Park BJ
Mon Apr 24 00:00:00 EDT 2023
Enamel defects of Axenfeld-Rieger syndrome and the role of PITX2 in its pathogenesis.
Oral diseases
Yang Y,Zhu J,Chiba Y,Fukumoto S,Qin M,Wang X
Wed Nov 01 00:00:00 EDT 2023
CircRNF10-DHX15 interaction suppressed breast cancer progression by antagonizing DHX15-NF-κB p65 positive feedback loop.
Cellular & molecular biology letters
Zheng W,Wang X,Yu Y,Ji C,Fang L
Wed Apr 26 00:00:00 EDT 2023
Whole exome sequencing reveals novel variants associated with diminished ovarian reserve in young women.
Frontiers in genetics
Li N,Xu W,Liu H,Zhou R,Zou S,Wang S,Li S,Yang Z,Piao Y,Zhang Y
Tue Apr 18 00:00:00 EDT 2023
The well-developed actin cytoskeleton and Cthrc1 expression by actin-binding protein drebrin in myofibroblasts promote cardiac and hepatic fibrosis.
The Journal of biological chemistry
Hironaka T,Takizawa N,Yamauchi Y,Horii Y,Nakaya M
Wed Mar 01 00:00:00 EST 2023
E2F1-mediated ectopic expression of PP1A promotes breast cancer progression via activation of YAP1.
The international journal of biochemistry & cell biology
Deng X,Hua K,Munankarmy A,Luo Q,Wang X,Fang L
Sat Apr 01 00:00:00 EDT 2023
Cerium Oxide Nanoparticles Promote Osteoplastic Precursor Differentiation by Activating the Wnt Pathway.
Biological trace element research
Luo J,Zhu S,Tong Y,Zhang Y,Li Y,Cao L,Kong M,Luo M,Bi Q,Zhang Q
Wed Feb 01 00:00:00 EST 2023
High expression of peroxisomal D-bifunctional protein in cytosol regulates apoptosis and energy metabolism of hepatocellular carcinoma cells via PI3K/AKT pathway.
American journal of cancer research
Zhang N,Wang YQ,Sun C,Shi Y,Hou LG,Yao M,Hu M,Wang XM,Ma PP,Li WJ,Jiang LL
Mon Jun 12 00:00:00 EDT 2023
Nogo-B deficiency suppresses white adipogenesis by regulating β-catenin signaling.
Life sciences
Li J,Sun Y,Xue C,Yang X,Duan Y,Zhao D,Han J
Mon May 15 00:00:00 EDT 2023
USP28 controls SREBP2 and the mevalonate pathway to drive tumour growth in squamous cancer.
Cell death and differentiation
Maier CR,Hartmann O,Prieto-Garcia C,Al-Shami KM,Schlicker L,Vogel FCE,Haid S,Klann K,Buck V,Münch C,Schmitz W,Einig E,Krenz B,Calzado MA,Eilers M,Popov N,Rosenfeldt MT,Diefenbacher ME,Schulze A
Sat Jul 01 00:00:00 EDT 2023
Characterization of huntingtin interactomes and their dynamic responses in living cells by proximity proteomics.
Journal of neurochemistry
Xu H,Bensalel J,Raju S,Capobianco E,Lu ML,Wei J
Wed Feb 01 00:00:00 EST 2023
RICH1 inhibits breast cancer stem cell traits through activating kinases cascade of Hippo signaling by competing with Merlin for binding to Amot-p80.
Cell death & disease
Tian Q,Gao H,Zhou Y,Zhu L,Yang J,Wang B,Liu P,Yang J
Fri Jan 21 00:00:00 EST 2022
p62 Promotes the Mitochondrial Localization of p53 through Its UBA Domain and Participates in Regulating the Sensitivity of Ovarian Cancer Cells to Cisplatin.
International journal of molecular sciences
Kong Q,Yan X,Cheng M,Jiang X,Xu L,Shen L,Yu H,Sun L
Fri Mar 18 00:00:00 EDT 2022
Tetrahydroxy Stilbene Glucoside Alleviates Ischemic Stroke by Regulating Conformation-Dependent Intracellular Distribution of PKM2 for M2 Macrophage Polarization.
Journal of agricultural and food chemistry
Li M,Lu W,Meng Y,Zhang W,Wang F,Sun L,Xu Y
Wed Dec 14 00:00:00 EST 2022
The perinuclear theca protein Calicin helps shape the sperm head and maintain the nuclear structure in mice.
Cell reports
Zhang XZ,Wei LL,Jin HJ,Zhang XH,Chen SR
Tue Jul 05 00:00:00 EDT 2022
MCM2 promotes the stemness and sorafenib resistance of hepatocellular carcinoma cells via hippo signaling.
Cell death discovery
Zhou X,Luo J,Xie H,Wei Z,Li T,Liu J,Liao X,Zhu G,Peng T
Sat Oct 15 00:00:00 EDT 2022
Multiple PDE3A modulators act as molecular glues promoting PDE3A-SLFN12 interaction and induce SLFN12 dephosphorylation and cell death.
Cell chemical biology
Yan B,Ding Z,Zhang W,Cai G,Han H,Ma Y,Cao Y,Wang J,Chen S,Ai Y
Thu Jun 16 00:00:00 EDT 2022
HDAC9 Contributes to Serous Ovarian Cancer Progression through Regulating Epithelial-Mesenchymal Transition.
Biomedicines
Xu L,Wang J,Liu B,Fu J,Zhao Y,Yu S,Shen L,Yan X,Su J
Thu Feb 03 00:00:00 EST 2022
Loss of perinuclear theca ACTRT1 causes acrosome detachment and severe male subfertility in mice.
Development (Cambridge, England)
Zhang XZ,Wei LL,Zhang XH,Jin HJ,Chen SR
Wed Jun 15 00:00:00 EDT 2022
Activation of Nrf2 modulates protective immunity against Mycobacterium tuberculosis infection in THP1-derived macrophages.
Free radical biology & medicine
Zhou J,Fang F,Qi J,Li T,Zhang L,Liu H,Lv J,Xu T,Wu F,Song C,Li W,Wang X,Chang X,Wang H,Wang T,Qian Z
Sun Nov 20 00:00:00 EST 2022
Irradiation combined with PD-L1-/- and autophagy inhibition enhances the antitumor effect of lung cancer via cGAS-STING-mediated T cell activation.
iScience
Zhao X,Hu S,Zeng L,Liu X,Song Y,Zhang Y,Chen Q,Bai Y,Zhang J,Zhang H,Pan Y,Shao C
Fri Aug 19 00:00:00 EDT 2022
Global crotonylome reveals hypoxia-mediated lamin A crotonylation regulated by HDAC6 in liver cancer.
Cell death & disease
Zhang D,Tang J,Xu Y,Huang X,Wang Y,Jin X,Wu G,Liu P
Wed Aug 17 00:00:00 EDT 2022
Global crotonylome reveals hypoxia-mediated lamin A crotonylation regulated by HDAC6 in liver cancer.
Cell death & disease
Zhang D,Tang J,Xu Y,Huang X,Wang Y,Jin X,Wu G,Liu P
Wed Aug 17 00:00:00 EDT 2022
Loss of Mature Lamin A/C Triggers a Shift in Intracellular Metabolic Homeostasis via AMPKα Activation.
Cells
Zhou Y,Yang JJ,Cheng Y,Feng GX,Yang RH,Yuan Y,Wang LY,Wang M,Kong L
Fri Dec 09 00:00:00 EST 2022
Loss of Mature Lamin A/C Triggers a Shift in Intracellular Metabolic Homeostasis via AMPKα Activation.
Cells
Zhou Y,Yang JJ,Cheng Y,Feng GX,Yang RH,Yuan Y,Wang LY,Wang M,Kong L
Fri Dec 09 00:00:00 EST 2022
Loss of Mature Lamin A/C Triggers a Shift in Intracellular Metabolic Homeostasis via AMPKα Activation.
Cells
Zhou Y,Yang JJ,Cheng Y,Feng GX,Yang RH,Yuan Y,Wang LY,Wang M,Kong L
Fri Dec 09 00:00:00 EST 2022
Loss of Mature Lamin A/C Triggers a Shift in Intracellular Metabolic Homeostasis via AMPKα Activation.
Cells
Zhou Y,Yang JJ,Cheng Y,Feng GX,Yang RH,Yuan Y,Wang LY,Wang M,Kong L
Fri Dec 09 00:00:00 EST 2022
Loss of Mature Lamin A/C Triggers a Shift in Intracellular Metabolic Homeostasis via AMPKα Activation.
Cells
Zhou Y,Yang JJ,Cheng Y,Feng GX,Yang RH,Yuan Y,Wang LY,Wang M,Kong L
Fri Dec 09 00:00:00 EST 2022
Loss of Mature Lamin A/C Triggers a Shift in Intracellular Metabolic Homeostasis via AMPKα Activation.
Cells
Zhou Y,Yang JJ,Cheng Y,Feng GX,Yang RH,Yuan Y,Wang LY,Wang M,Kong L
Fri Dec 09 00:00:00 EST 2022
AKR1B10 accelerates the production of proinflammatory cytokines via the NF-κB signaling pathway in colon cancer.
Journal of molecular histology
Liu C,Shi L,Li W,Huang Z,Wang S,Xu P,Li T,Li Z,Luo F,Li W,Yan J,Wu T
Sat Oct 01 00:00:00 EDT 2022
Qushi Huayu decoction attenuated hepatic lipid accumulation via JAK2/STAT3/CPT-1A-related fatty acid β-oxidation in mice with non-alcoholic steatohepatitis.
Pharmaceutical biology
Sun Q,Wang X,Xin X,An Z,Hu Y,Feng Q
Thu Dec 01 00:00:00 EST 2022
Nrf2 Overexpression Decreases Vincristine Chemotherapy Sensitivity Through the PI3K-AKT Pathway in Adult B-Cell Acute Lymphoblastic Leukemia.
Frontiers in oncology
Wang L,Liu X,Kang Q,Pan C,Zhang T,Feng C,Chen L,Wei S,Wang J
Sat Jul 16 00:00:00 EDT 2022
MITF activity is regulated by a direct interaction with RAF proteins in melanoma cells.
Communications biology
Estrada C,Mirabal-Ortega L,Méry L,Dingli F,Besse L,Messaoudi C,Loew D,Pouponnot C,Bertolotto C,Eychène A,Druillennec S
Fri Jan 28 00:00:00 EST 2022
Suberoylanilide Hydroxamic Acid (SAHA) Treatment Reveals Crosstalk Among Proteome, Phosphoproteome, and Acetylome in Nasopharyngeal Carcinoma Cells.
Frontiers in genetics
Huang H,Fu Y,Duan Y,Zhang Y,Lu M,Chen Z,Li M,Chen Y
Sat Jul 16 00:00:00 EDT 2022
Suberoylanilide Hydroxamic Acid (SAHA) Treatment Reveals Crosstalk Among Proteome, Phosphoproteome, and Acetylome in Nasopharyngeal Carcinoma Cells.
Frontiers in genetics
Huang H,Fu Y,Duan Y,Zhang Y,Lu M,Chen Z,Li M,Chen Y
Sat Jul 16 00:00:00 EDT 2022
miR-376a-3p and miR-376b-3p overexpression in Hutchinson-Gilford progeria fibroblasts inhibits cell proliferation and induces premature senescence.
iScience
Frankel D,Delecourt V,Novoa-Del-Toro EM,Robin JD,Airault C,Bartoli C,Carabalona A,Perrin S,Mazaleyrat K,De Sandre-Giovannoli A,Magdinier F,Baudot A,Lévy N,Kaspi E,Roll P
Fri Feb 18 00:00:00 EST 2022
miR-376a-3p and miR-376b-3p overexpression in Hutchinson-Gilford progeria fibroblasts inhibits cell proliferation and induces premature senescence.
iScience
Frankel D,Delecourt V,Novoa-Del-Toro EM,Robin JD,Airault C,Bartoli C,Carabalona A,Perrin S,Mazaleyrat K,De Sandre-Giovannoli A,Magdinier F,Baudot A,Lévy N,Kaspi E,Roll P
Fri Feb 18 00:00:00 EST 2022
Keratin 17 Promotes T Cell Response in Allergic Contact Dermatitis by Upregulating C-C Motif Chemokine Ligand 20.
Frontiers in immunology
Luo Y,Zhu Z,Li B,Bai X,Fang H,Qiao P,Chen J,Zhang C,Zhi D,Dang E,Wang G
Wed Mar 02 00:00:00 EST 2022
Highly efficient A-to-G base editing by ABE8.17 in rabbits.
Molecular therapy. Nucleic acids
Zhao D,Qian Y,Li J,Li Z,Lai L
Tue Mar 08 00:00:00 EST 2022
Inhibition of FOXO1‑mediated autophagy promotes paclitaxel‑induced apoptosis of MDA‑MB‑231 cells.
Molecular medicine reports
Xu K,Zhu W,Xu A,Xiong Z,Zou D,Zhao H,Jiao D,Qing Y,Jamal MA,Wei HJ,Zhao HY
Tue Feb 01 00:00:00 EST 2022
Targeting epiregulin in the treatment-damaged tumor microenvironment restrains therapeutic resistance.
Oncogene
Wang C,Long Q,Fu Q,Xu Q,Fu D,Li Y,Gao L,Guo J,Zhang X,Lam EW,Campisi J,Sun Y
Tue Nov 01 00:00:00 EDT 2022
Synaptic proteomics reveal distinct molecular signatures of cognitive change and C9ORF72 repeat expansion in the human ALS cortex.
Acta neuropathologica communications
Laszlo ZI,Hindley N,Sanchez Avila A,Kline RA,Eaton SL,Lamont DJ,Smith C,Spires-Jones TL,Wishart TM,Henstridge CM
Sat Oct 29 00:00:00 EDT 2022
Cell-Penetrating Peptide TAT-HuR-HNS3 Suppresses Proinflammatory Gene Expression via Competitively Blocking Interaction of HuR with Its Partners.
Journal of immunology (Baltimore, Md. : 1950)
Wang K,Tong H,Gao Y,Xia L,Jin X,Li X,Zeng X,Boldogh I,Ke Y,Ba X
Sun May 15 00:00:00 EDT 2022
Caspase-1-mediated extracellular vesicles derived from pyroptotic alveolar macrophages promote inflammation in acute lung injury.
International journal of biological sciences
Qin X,Zhou Y,Jia C,Chao Z,Qin H,Liang J,Liu X,Liu Z,Sun T,Yuan Y,Zhang H
Fri Apr 15 00:00:00 EDT 2022
Substrate rigidity dictates colorectal tumorigenic cell stemness and metastasis via CRAD-dependent mechanotransduction.
Cell reports
Chang Y,Zhang J,Huo X,Qu X,Xia C,Huang K,Xie F,Wang N,Wei X,Jia Q
Tue Feb 15 00:00:00 EST 2022
SETDB1 fuels the lung cancer phenotype by modulating epigenome, 3D genome organization and chromatin mechanical properties.
Nucleic acids research
Zakharova VV,Magnitov MD,Del Maestro L,Ulianov SV,Glentis A,Uyanik B,Williart A,Karpukhina A,Demidov O,Joliot V,Vassetzky YS,Mège RM,Piel M,Razin SV,Ait-Si-Ali S
Fri May 06 00:00:00 EDT 2022
Inhibition of high-fat diet-induced obesity via reduction of ER-resident protein Nogo occurs through multiple mechanisms.
The Journal of biological chemistry
Wang X,Yang Y,Zhao D,Zhang S,Chen Y,Chen Y,Feng K,Li X,Han J,Iwakiri Y,Duan Y,Yang X
Tue Feb 01 00:00:00 EST 2022
Disturbing NLRP3 acetylation and inflammasome assembly inhibits androgen receptor-promoted inflammatory responses and prostate cancer progression.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Zhao AN,Yang Z,Wang DD,Shi B,Zhang H,Bai Y,Yan BW,Zhang Y,Wen JK,Wang XL,Qu CB
Tue Nov 01 00:00:00 EDT 2022
Mouse Ocilrp2/Clec2i negatively regulates LPS-mediated IL-6 production by blocking Dap12-Syk interaction in macrophage.
Frontiers in immunology
Cao M,Ma L,Yan C,Wang H,Ran M,Chen Y,Wang X,Liang X,Chai L,Li X
Fri Oct 28 00:00:00 EDT 2022
Lactylation-driven METTL3-mediated RNA m6A modification promotes immunosuppression of tumor-infiltrating myeloid cells.
Molecular cell
Xiong J,He J,Zhu J,Pan J,Liao W,Ye H,Wang H,Song Y,Du Y,Cui B,Xue M,Zheng W,Kong X,Jiang K,Ding K,Lai L,Wang Q
Thu May 05 00:00:00 EDT 2022
A novel homozygous missense mutation in AK7 causes multiple morphological anomalies of the flagella and oligoasthenoteratozoospermia.
Journal of assisted reproduction and genetics
Xiang M,Wang Y,Xu W,Zheng N,Deng H,Zhang J,Duan Z,Zha X,Zhang W,Song G,Shi X,Wang F,Cao Y,Zhu F
Sat Jan 01 00:00:00 EST 2022
Metformin Ameliorates Chronic Colitis-Related Intestinal Fibrosis via Inhibiting TGF-β1/Smad3 Signaling.
Frontiers in pharmacology
Wang Y,Wang Z,Yang H,Chen S,Zheng D,Liu X,Jiang Q,Chen Y
Sat Jul 16 00:00:00 EDT 2022
Novel Mutation and Deletion in SUN5 Cause Male Infertility with Acephalic Spermatozoa Syndrome.
Reproductive sciences (Thousand Oaks, Calif.)
Xiang M,Wang Y,Wang K,Kong S,Lu M,Zhang J,Duan Z,Zha X,Shi X,Wang F,Cao Y,Zhu F
Tue Feb 01 00:00:00 EST 2022
Metabolomics of clinical samples reveal the treatment mechanism of lanthanum hydroxide on vascular calcification in chronic kidney disease.
Proceedings of the Japan Academy. Series B, Physical and biological sciences
Gu C,Gao Y,Han R,Guo M,Liu H,Gao J,Liu Y,Li B,Sun L,Bu R,Liu Y,Hao J,Meng Y,An M,Cao X,Su C,Li G
Tue Aug 02 00:00:00 EDT 2022
BRCA2 promoter hypermethylation as a biomarker for the leukemic transformation of myeloproliferative neoplasms.
Epigenomics
Yang C,Zhang Q,Tang X,Wang B,Guan M,Tang G,Wu Z
Fri Apr 01 00:00:00 EDT 2022
Static Magnetic Fields Reduce Oxidative Stress to Improve Wound Healing and Alleviate Diabetic Complications.
Cells
Feng C,Yu B,Song C,Wang J,Zhang L,Ji X,Wang Y,Fang Y,Liao Z,Wei M,Zhang X
Thu Jan 27 00:00:00 EST 2022
Hsa_circ_0005050 interacts with ILF3 and affects cell apoptosis and proliferation by disrupting the balance between p53 and p65.
Chemico-biological interactions
Tan J,Sun M,Yin J,Zhou Q,Zhao R,Chen Q,Sun H,Jiang C,Li S,He Y
Thu Dec 01 00:00:00 EST 2022
Lanthanum hydroxide inhibits vascular calcification by regulating the HIF-1 pathway.
Cell biology international
Wang S,Zhao L,Liu H,Han R,Liu Y,Gao J,Bu R,Li B,Sun L,Hao J,Zhao J,Li G
Mon Aug 01 00:00:00 EDT 2022
Deletion of Smurf1 attenuates liver steatosis via stabilization of p53.
Laboratory investigation; a journal of technical methods and pathology
Lin W,Zhang X,Zhang C,Li L,Zhang J,Xie P,Zhan Y,An W
Sat Oct 01 00:00:00 EDT 2022
Olfactory marker protein contains a leucine-rich domain in the Ω-loop important for nuclear export.
Molecular brain
Nakashima N,Nakashima A,Nakashima K,Takano M
Fri Nov 04 00:00:00 EDT 2022
Cimigenoside functions as a novel γ-secretase inhibitor and inhibits the proliferation or metastasis of human breast cancer cells by γ-secretase/Notch axis.
Pharmacological research
Jia H,Liu M,Wang X,Jiang Q,Wang S,Santhanam RK,Lv C,Zhao Q,Lu J
Thu Jul 01 00:00:00 EDT 2021
Cryptotanshinone, a novel PDK 4 inhibitor, suppresses bladder cancer cell invasiveness via the mTOR/β‑catenin/N‑cadherin axis.
International journal of oncology
Kim CJ,Terado T,Tambe Y,Mukaisho KI,Kageyama S,Kawauchi A,Inoue H
Thu Jul 01 00:00:00 EDT 2021
Inducible ATP1B1 Upregulates Antiviral Innate Immune Responses by the Ubiquitination of TRAF3 and TRAF6.
Journal of immunology (Baltimore, Md. : 1950)
Cao W,Guo Y,Cheng Z,Xu G,Zuo Q,Nie L,Huang Y,Liu S,Zhu Y
Tue Jun 01 00:00:00 EDT 2021
Long non-coding RNA NR2F1-AS1 induces breast cancer lung metastatic dormancy by regulating NR2F1 and ΔNp63.
Nature communications
Liu Y,Zhang P,Wu Q,Fang H,Wang Y,Xiao Y,Cong M,Wang T,He Y,Ma C,Tian P,Liang Y,Qin LX,Yang Q,Yang Q,Liao L,Hu G
Thu Sep 02 00:00:00 EDT 2021
Arsenic trioxide induces expression of BCL-2 expression via NF-κB and p38 MAPK signaling pathways in BEAS-2B cells during apoptosis.
Ecotoxicology and environmental safety
Tang J,Yao C,Liu Y,Yuan J,Wu L,Hosoi K,Yu S,Huang C,Wei H,Chen G
Fri Oct 01 00:00:00 EDT 2021
HSD17B6 downregulation predicts poor prognosis and drives tumor progression via activating Akt signaling pathway in lung adenocarcinoma.
Cell death discovery
Tian T,Hong F,Wang Z,Hu J,Chen N,Lv L,Yi Q
Mon Nov 08 00:00:00 EST 2021
Nuclear Factor-κB Signaling Mediates Antimony-induced Astrocyte Activation.
Biomedical and environmental sciences : BES
Zhang T,Zheng YD,Jiao M,Zhi Y,Xu SY,Zhu PY,Zhao XY,Wu QY
Wed Jan 20 00:00:00 EST 2021
Formin-like protein 2 promotes cell proliferation by a p27-related mechanism in human breast cancer cells.
BMC cancer
Jiao X,Wang B,Feng C,Song S,Tian B,Zhou C,Gao X,Sun W,Liu P
Wed Jun 30 00:00:00 EDT 2021
miR-1258 Attenuates Tumorigenesis Through Targeting E2F1 to Inhibit PCNA and MMP2 Transcription in Glioblastoma.
Frontiers in oncology
Qin H,Gui Y,Ma R,Zhang H,Guo Y,Ye Y,Li J,Zhao L,Wang Y
Sat Jun 05 00:00:00 EDT 2021
Prenylated xanthones from mangosteen (Garcinia mangostana) activate the AhR and Nrf2 pathways and protect intestinal barrier integrity in HT-29 cells.
Free radical biology & medicine
Tocmo R,Le B,Heun A,van Pijkeren JP,Parkin K,Johnson JJ
Mon Feb 01 00:00:00 EST 2021
Folic Acid Protects Melanocytes from Oxidative Stress via Activation of Nrf2 and Inhibition of HMGB1.
Oxidative medicine and cellular longevity
Du P,Zhang S,Li S,Yang Y,Kang P,Chen J,Gao T,Li C,Zhang Q,Zhang W
Mon Feb 28 00:00:00 EST 2022
The Pharmacological Effect and Mechanism of Lanthanum Hydroxide on Vascular Calcification Caused by Chronic Renal Failure Hyperphosphatemia.
Frontiers in cell and developmental biology
Zhao L,Wang S,Liu H,Du X,Bu R,Li B,Han R,Gao J,Liu Y,Hao J,Zhao J,Meng Y,Li G
Fri Apr 22 00:00:00 EDT 2022
hsa_circ_0068631 promotes breast cancer progression through c-Myc by binding to EIF4A3.
Molecular therapy. Nucleic acids
Wang X,Chen M,Fang L
Fri Dec 03 00:00:00 EST 2021
Virus-inducible IGFALS facilitates innate immune responses by mediating IRAK1 and TRAF6 activation.
Cellular & molecular immunology
Xu G,Deng F,Zuo Q,Liu L,Dou K,Cheng Z,Cao W,Luo C,Yu C,Liu S,Zhu Y
Tue Jun 01 00:00:00 EDT 2021
Autophagy-Induced HDAC6 Activity During Hypoxia Regulates Mitochondrial Energy Metabolism Through the β-Catenin/COUP-TFII Axis in Hepatocellular Carcinoma Cells.
Frontiers in oncology
Yan X,Qu X,Liu B,Zhao Y,Xu L,Yu S,Wang J,Wang L,Su J
Tue Dec 07 00:00:00 EST 2021
The role of LR-TIMAP/PP1c complex in the occurrence and development of no-reflow.
EBioMedicine
Quan X,Liu X,Qin X,Wang Y,Sun T,Li Z,Zhu L,Chen J,Zhou Y,Singh S,Dong H,Zhang Z,Zhang H
Mon Mar 01 00:00:00 EST 2021
Extracellular vesicles derived from oesophageal cancer containing P4HB promote muscle wasting via regulating PHGDH/Bcl-2/caspase-3 pathway.
Journal of extracellular vesicles
Gao X,Wang Y,Lu F,Chen X,Yang D,Cao Y,Zhang W,Chen J,Zheng L,Wang G,Fu M,Ma L,Song Y,Zhan Q
Mon Mar 01 00:00:00 EST 2021
Hsa_circ_0005273 facilitates breast cancer tumorigenesis by regulating YAP1-hippo signaling pathway.
Journal of experimental & clinical cancer research : CR
Wang X,Ji C,Hu J,Deng X,Zheng W,Yu Y,Hua K,Zhou X,Fang L
Tue Jan 12 00:00:00 EST 2021
The Retinoblastoma Tumor Suppressor Is Required for the NUP98-HOXA9-Induced Aberrant Nuclear Envelope Phenotype.
Cells
Vaz M,Fahrenkrog B
Fri Oct 22 00:00:00 EDT 2021
Adipocyte-Specific Deletion of Lamin A/C Largely Models Human Familial Partial Lipodystrophy Type 2.
Diabetes
Corsa CAS,Walsh CM,Bagchi DP,Foss Freitas MC,Li Z,Hardij J,Granger K,Mori H,Schill RL,Lewis KT,Maung JN,Azaria RD,Rothberg AE,Oral EA,MacDougald OA
Wed Sep 01 00:00:00 EDT 2021
AKR1B10 negatively regulates autophagy through reducing GAPDH upon glucose starvation in colon cancer.
Journal of cell science
Li W,Liu C,Huang Z,Shi L,Zhong C,Zhou W,Meng P,Li Z,Wang S,Luo F,Yan J,Wu T
Thu Apr 15 00:00:00 EDT 2021
Role of IL-6-IL-27 Complex in Host Antiviral Immune Response.
Journal of immunology (Baltimore, Md. : 1950)
Zuo Q,Cheng Z,Zhang G,Xia Y,Xu G,Cao W,Yang X,Fu Y,He R,Fang P,Guo Y,Nie L,Huang Y,Liu L,Zhan J,Liu S,Zhu Y
Thu Jul 15 00:00:00 EDT 2021
The flavonoid procyanidin C1 has senotherapeutic activity and increases lifespan in mice.
Nature metabolism
Xu Q,Fu Q,Li Z,Liu H,Wang Y,Lin X,He R,Zhang X,Ju Z,Campisi J,Kirkland JL,Sun Y
Wed Dec 01 00:00:00 EST 2021
Bortezomib potentiates antitumor activity of mitoxantrone through dampening Wnt/β-catenin signal pathway in prostate cancer cells.
BMC cancer
Zhang Y,Liu Q,Wei W,Zhang G,Yan S,Dai R,Sun Y,Su D,Lv S,Xia Y,Li J,Li C
Wed Oct 13 00:00:00 EDT 2021
The miR-378c-Samd1 circuit promotes phenotypic modulation of vascular smooth muscle cells and foam cells formation in atherosclerosis lesions.
Scientific reports
Tian S,Cao Y,Wang J,Bi Y,Zhong J,Meng X,Sun W,Yang R,Gan L,Wang X,Li H,Wang R
Tue May 18 00:00:00 EDT 2021
Syndecan 4 contributes to osteoclast differentiation induced by RANKL through enhancing autophagy.
International immunopharmacology
Li J,Sun Z,Lin Y,Yan Y,Yan H,Jing B,Han Z
Mon Feb 01 00:00:00 EST 2021
Hepatitis B Surface Antigen Suppresses the Activation of Nuclear Factor Kappa B Pathway via Interaction With the TAK1-TAB2 Complex.
Frontiers in immunology
Deng F,Xu G,Cheng Z,Huang Y,Ma C,Luo C,Yu C,Wang J,Xu X,Liu S,Zhu Y
Mon Jun 28 00:00:00 EDT 2021
EEF1A2 interacts with HSP90AB1 to promote lung adenocarcinoma metastasis via enhancing TGF-β/SMAD signalling.
British journal of cancer
Jia L,Ge X,Du C,Chen L,Zhou Y,Xiong W,Xiang J,Li G,Xiao G,Fang L,Li Z
Mon Mar 01 00:00:00 EST 2021
Carnosic acid protects against ferroptosis in PC12 cells exposed to erastin through activation of Nrf2 pathway.
Life sciences
Cheng J,Xu T,Xun C,Guo H,Cao R,Gao S,Sheng W
Mon Feb 01 00:00:00 EST 2021
Nuclear speckle specific hnRNP D-like prevents age- and AD-related cognitive decline by modulating RNA splicing.
Molecular neurodegeneration
Zhang Q,Zhang J,Ye J,Li X,Liu H,Ma X,Wang C,He K,Zhang W,Yuan J,Zhao Y,Xu H,Liu Q
Wed Sep 22 00:00:00 EDT 2021
Epiregulin (EREG) and Myocardin Related Transcription Factor A (MRTF-A) Form a Feedforward Loop to Drive Hepatic Stellate Cell Activation.
Frontiers in cell and developmental biology
Wu X,Dong W,Zhang T,Ren H,Wang J,Shang L,Zhu Z,Zhu W,Shi X,Xu Y
Tue Feb 02 00:00:00 EST 2021
HuR stabilizes HTT mRNA via interacting with its exon 11 in a mutant HTT-dependent manner.
RNA biology
Zhao Q,Li C,Yu M,Sun Y,Wang J,Ma L,Sun X,Lu B
Wed Apr 01 00:00:00 EDT 2020
p62/SQSTM1 accumulation due to degradation inhibition and transcriptional activation plays a critical role in silica nanoparticle-induced airway inflammation via NF-κB activation.
Journal of nanobiotechnology
Wu Y,Jin Y,Sun T,Zhu P,Li J,Zhang Q,Wang X,Jiang J,Chen G,Zhao X
Tue May 19 00:00:00 EDT 2020
Torsin ATPases influence chromatin interaction of the Torsin regulator LAP1.
eLife
Luithle N,de Bos JU,Hovius R,Maslennikova D,Lewis RT,Ungricht R,Fierz B,Kutay U
Tue Dec 15 00:00:00 EST 2020
SSEA3 and Sialyl Lewis a Glycan Expression Is Controlled by B3GALT5 LTR through Lamin A-NFYA and SIRT1-STAT3 Signaling in Human ES Cells.
Cells
Cai BH,Lee HY,Chou CK,Wu PH,Huang HC,Chao CC,Chung HY,Kannagi R
Fri Jan 10 00:00:00 EST 2020
SOX2 Represses Hepatitis B Virus Replication by Binding to the Viral EnhII/Cp and Inhibiting the Promoter Activation.
Viruses
Yang H,Mo J,Xiang Q,Zhao P,Song Y,Yang G,Wu K,Liu Y,Liu W,Wu J
Sat Feb 29 00:00:00 EST 2020
N-myc downstream-regulated gene 2 promotes the protein stability of estrogen receptor beta via inhibition of ubiquitin-protein ligase E3A to suppress colorectal cancer.
Journal of gastrointestinal oncology
Zhu J,Lv Y,Hao J,Shi T,Wang S,Wang K,Fan X,Guo Y,Zhang J,Li J
Tue Dec 01 00:00:00 EST 2020
Heme Oxygenase-1 Suppresses Wnt Signaling Pathway in Nonalcoholic Steatohepatitis-Related Liver Fibrosis.
BioMed research international
Du J,Ren W,Zhang Q,Fu N,Han F,Cui P,Li W,Kong L,Zhao S,Wang R,Zhang Y,Yang L,Kong L,Nan Y
Fri Mar 12 00:00:00 EST 2021
Eukaryotic Translation Elongation Factor 1 Delta Inhibits the Nuclear Import of the Nucleoprotein and PA-PB1 Heterodimer of Influenza A Virus.
Journal of virology
Gao Q,Yang C,Ren C,Zhang S,Gao X,Jin M,Chen H,Ma W,Zhou H
Tue Dec 22 00:00:00 EST 2020
The CDK inhibitor p57Kip2 enhances the activity of the transcriptional coactivator FHL2.
Scientific reports
Kullmann MK,Podmirseg SR,Roilo M,Hengst L
Tue Apr 28 00:00:00 EDT 2020
Nuclear pore blockade reveals that HIV-1 completes reverse transcription and uncoating in the nucleus.
Nature microbiology
Dharan A,Bachmann N,Talley S,Zwikelmaier V,Campbell EM
Tue Sep 01 00:00:00 EDT 2020
Single-cell RNA-sequencing analysis identifies host long noncoding RNA MAMDC2-AS1 as a co-factor for HSV-1 nuclear transport.
International journal of biological sciences
Wang Y,Huang L,Wang Y,Luo W,Li F,Xiao J,Qin S,Wang Z,Song X,Wang Y,Jin F,Wang Y
Mon Aug 23 00:00:00 EDT 2021
α-Ketoglutarate Modulates Macrophage Polarization Through Regulation of PPARγ Transcription and mTORC1/p70S6K Pathway to Ameliorate ALI/ARDS.
Shock (Augusta, Ga.)
Liu M,Chen Y,Wang S,Zhou H,Feng D,Wei J,Shi X,Wu L,Zhang P,Yang H,Lv X
Wed Jan 01 00:00:00 EST 2020
Hepatitis B Core Antigen Impairs the Polarization While Promoting the Production of Inflammatory Cytokines of M2 Macrophages via the TLR2 Pathway.
Frontiers in immunology
Yi H,Zhang Y,Yang X,Li M,Hu H,Xiong J,Wang N,Jin J,Zhang Y,Song Y,Wang X,Chen L,Lian J
Fri Mar 26 00:00:00 EDT 2021
Simvastatin re-sensitizes hepatocellular carcinoma cells to sorafenib by inhibiting HIF-1α/PPAR-γ/PKM2-mediated glycolysis.
Journal of experimental & clinical cancer research : CR
Feng J,Dai W,Mao Y,Wu L,Li J,Chen K,Yu Q,Kong R,Li S,Zhang J,Ji J,Wu J,Mo W,Xu X,Guo C
Thu Jan 30 00:00:00 EST 2020
Effects of mevalonate kinase interference on cell differentiation, apoptosis, prenylation and geranylgeranylation of human keratinocytes are attenuated by farnesyl pyrophosphate or geranylgeranyl pyrophosphate.
Experimental and therapeutic medicine
Li M,Min W,Wang J,Wang L,Li Y,Zhou N,Yang Z,Qian Q
Wed Apr 01 00:00:00 EDT 2020
SARS-CoV-2 Nucleocapsid Protein Interacts with RIG-I and Represses RIG-Mediated IFN-β Production.
Viruses
Chen K,Xiao F,Hu D,Ge W,Tian M,Wang W,Pan P,Wu K,Wu J
Wed Dec 30 00:00:00 EST 2020
LL-37 restored glucocorticoid sensitivity impaired by virus dsRNA in lung.
International immunopharmacology
Li K,Tao N,Zheng L,Sun T
Sat Feb 01 00:00:00 EST 2020
Translation Efficiency and Degradation of ER-Associated mRNAs Modulated by ER-Anchored poly(A)-Specific Ribonuclease (PARN).
Cells
Duan TL,Jiao H,He GJ,Yan YB
Thu Jan 09 00:00:00 EST 2020
Tissue-Resident Alveolar Macrophages Do Not Rely on Glycolysis for LPS-induced Inflammation.
American journal of respiratory cell and molecular biology
Woods PS,Kimmig LM,Meliton AY,Sun KA,Tian Y,O'Leary EM,Gökalp GA,Hamanaka RB,Mutlu GM
Sat Feb 01 00:00:00 EST 2020
Osteopetrosis-Associated Transmembrane Protein 1 Recruits RNA Exosome To Restrict Hepatitis B Virus Replication.
Journal of virology
Ma C,Xu W,Yang Q,Liu W,Xiang Q,Chen J,Zhang Q,Liu Y,Wu K,Wu J
Mon May 18 00:00:00 EDT 2020
Cytoplasmic PCNA is located in the actin belt and involved in osteoclast differentiation.
Aging
Tang D,Liu X,Chen K,Li Z,Dai Y,Xu J,Zhang HT,Gao X,Liu L
Sat Jun 27 00:00:00 EDT 2020
Inhibition of protein phosphatase methylesterase 1 dysregulates MAP kinase signaling and attenuates muscle cell differentiation.
Gene
Labuzan SA,Lynch SA,Cooper LM,Waddell DS
Wed May 20 00:00:00 EDT 2020
Overexpression of NRF1-742 or NRF1-772 Reduces Arsenic-Induced Cytotoxicity and Apoptosis in Human HaCaT Keratinocytes.
International journal of molecular sciences
Wang S,Cheng H,Wang L,Zhao R,Guan D
Mon Mar 16 00:00:00 EDT 2020
Interplay of m6A and H3K27 trimethylation restrains inflammation during bacterial infection.
Science advances
Wu C,Chen W,He J,Jin S,Liu Y,Yi Y,Gao Z,Yang J,Yang J,Cui J,Zhao W
Sat Aug 01 00:00:00 EDT 2020
HSPA12A Is a Novel Player in Nonalcoholic Steatohepatitis via Promoting Nuclear PKM2-Mediated M1 Macrophage Polarization.
Diabetes
Kong Q,Li N,Cheng H,Zhang X,Cao X,Qi T,Dai L,Zhang Z,Chen X,Li C,Li Y,Xue B,Fang L,Liu L,Ding Z
Fri Feb 01 00:00:00 EST 2019
HSPA12A Is a Novel Player in Nonalcoholic Steatohepatitis via Promoting Nuclear PKM2-Mediated M1 Macrophage Polarization.
Diabetes
Kong Q,Li N,Cheng H,Zhang X,Cao X,Qi T,Dai L,Zhang Z,Chen X,Li C,Li Y,Xue B,Fang L,Liu L,Ding Z
Fri Feb 01 00:00:00 EST 2019
Lamin A mutation impairs interaction with nucleoporin NUP155 and disrupts nucleocytoplasmic transport in atrial fibrillation.
Human mutation
Han M,Zhao M,Cheng C,Huang Y,Han S,Li W,Tu X,Luo X,Yu X,Liu Y,Chen Q,Ren X,Wang QK,Ke T
Fri Mar 01 00:00:00 EST 2019
Lamin A mutation impairs interaction with nucleoporin NUP155 and disrupts nucleocytoplasmic transport in atrial fibrillation.
Human mutation
Han M,Zhao M,Cheng C,Huang Y,Han S,Li W,Tu X,Luo X,Yu X,Liu Y,Chen Q,Ren X,Wang QK,Ke T
Fri Mar 01 00:00:00 EST 2019
Matrix stiffness mediates stemness characteristics via activating the Yes-associated protein in colorectal cancer cells.
Journal of cellular biochemistry
Tan F,Huang Y,Pei Q,Liu H,Pei H,Zhu H
Fri Feb 01 00:00:00 EST 2019
Matrix stiffness mediates stemness characteristics via activating the Yes-associated protein in colorectal cancer cells.
Journal of cellular biochemistry
Tan F,Huang Y,Pei Q,Liu H,Pei H,Zhu H
Fri Feb 01 00:00:00 EST 2019
Inducible LGALS3BP/90K activates antiviral innate immune responses by targeting TRAF6 and TRAF3 complex.
PLoS pathogens
Xu G,Xia Z,Deng F,Liu L,Wang Q,Yu Y,Wang F,Zhu C,Liu W,Cheng Z,Zhu Y,Zhou L,Zhang Y,Lu M,Liu S
Thu Aug 01 00:00:00 EDT 2019
HSF1 phosphorylation by cyclosporin A confers hyperthermia sensitivity through suppression of HSP expression.
Biochimica et biophysica acta. Gene regulatory mechanisms
Shao J,Han B,Cao P,Zhang B,Liu M,Li D,Zhou N,Hao Q,Duan X,Chang Y,Nakai A,Fan Y,Tan K
Thu Aug 01 00:00:00 EDT 2019
PKM2 is the target of proanthocyanidin B2 during the inhibition of hepatocellular carcinoma.
Journal of experimental & clinical cancer research : CR
Feng J,Wu L,Ji J,Chen K,Yu Q,Zhang J,Chen J,Mao Y,Wang F,Dai W,Xu L,Wu J,Guo C
Fri May 17 00:00:00 EDT 2019
Angiomotin-p130 inhibits β-catenin stability by competing with Axin for binding to tankyrase in breast cancer.
Cell death & disease
Yang J,Zhang X,Chen Z,Shen Y,Wang F,Wang Y,Liu Y,Liu P,Yang J
Thu Feb 21 00:00:00 EST 2019
MiR-27a-regulated FOXO1 promotes pancreatic ductal adenocarcinoma cell progression by enhancing Wnt/β-catenin signaling activity.
American journal of translational research
Ling J,Dong X,Wang L,Xue Y,Jia X,Song W,Li Q
Tue Sep 29 00:00:00 EDT 2020
NAC1 Potentiates Cellular Antiviral Signaling by Bridging MAVS and TBK1.
Journal of immunology (Baltimore, Md. : 1950)
Xia Z,Xu G,Nie L,Liu L,Peng N,He Q,Zuo Q,Zhou Y,Cao Z,Liu S,Zhu Y
Thu Aug 15 00:00:00 EDT 2019
JNK activation-mediated nuclear SIRT1 protein suppression contributes to silica nanoparticle-induced pulmonary damage via p53 acetylation and cytoplasmic localisation.
Toxicology
Zhao X,Wu Y,Li J,Li D,Jin Y,Zhu P,Liu Y,Zhuang Y,Yu S,Cao W,Wei H,Wang X,Han Y,Chen G
Mon Jul 01 00:00:00 EDT 2019
YAP mediates the positive regulation of hnRNPK on the lung adenocarcinoma H1299 cell growth.
Acta biochimica et biophysica Sinica
Xu L,Zhang T,Huang W,Liu X,Lu J,Gao X,Zhang YF,Liu L
Wed Jul 10 00:00:00 EDT 2019
miR‑494‑3p regulates lipopolysaccharide‑induced inflammatory responses in RAW264.7 cells by targeting PTEN.
Molecular medicine reports
Zhang S,He K,Zhou W,Cao J,Jin Z
Wed May 01 00:00:00 EDT 2019
CRABP2 regulates invasion and metastasis of breast cancer through hippo pathway dependent on ER status.
Journal of experimental & clinical cancer research : CR
Feng X,Zhang M,Wang B,Zhou C,Mu Y,Li J,Liu X,Wang Y,Song Z,Liu P
Fri Aug 16 00:00:00 EDT 2019
Inhibition of Influenza A Virus Replication by TRIM14 via Its Multifaceted Protein-Protein Interaction With NP.
Frontiers in microbiology
Wu X,Wang J,Wang S,Wu F,Chen Z,Li C,Cheng G,Qin FX
Thu Oct 01 00:00:00 EDT 2020
Deficiency of heat shock protein A12A promotes browning of white adipose tissues in mice.
Biochimica et biophysica acta. Molecular basis of disease
Cheng H,Qi T,Zhang X,Kong Q,Min X,Mao Q,Cao X,Liu L,Ding Z
Sat Jun 01 00:00:00 EDT 2019
SET1A-Mediated Mono-Methylation at K342 Regulates YAP Activation by Blocking Its Nuclear Export and Promotes Tumorigenesis.
Cancer cell
Fang L,Teng H,Wang Y,Liao G,Weng L,Li Y,Wang X,Jin J,Jiao C,Chen L,Peng X,Chen J,Yang Y,Fang H,Han D,Li C,Jin X,Zhang S,Liu Z,Liu M,Wei Q,Liao L,Ge X,Zhao B,Zhou D,Qin HL,Zhou J,Wang P
Mon Jul 09 00:00:00 EDT 2018
SET1A-Mediated Mono-Methylation at K342 Regulates YAP Activation by Blocking Its Nuclear Export and Promotes Tumorigenesis.
Cancer cell
Fang L,Teng H,Wang Y,Liao G,Weng L,Li Y,Wang X,Jin J,Jiao C,Chen L,Peng X,Chen J,Yang Y,Fang H,Han D,Li C,Jin X,Zhang S,Liu Z,Liu M,Wei Q,Liao L,Ge X,Zhao B,Zhou D,Qin HL,Zhou J,Wang P
Mon Jul 09 00:00:00 EDT 2018
Hypoxia-Induced Mitogenic Factor Promotes Cardiac Hypertrophy via Calcium-Dependent and Hypoxia-Inducible Factor-1α Mechanisms.
Hypertension (Dallas, Tex. : 1979)
Kumar S,Wang G,Liu W,Ding W,Dong M,Zheng N,Ye H,Liu J
Wed Aug 01 00:00:00 EDT 2018
Hypoxia-Induced Mitogenic Factor Promotes Cardiac Hypertrophy via Calcium-Dependent and Hypoxia-Inducible Factor-1α Mechanisms.
Hypertension (Dallas, Tex. : 1979)
Kumar S,Wang G,Liu W,Ding W,Dong M,Zheng N,Ye H,Liu J
Wed Aug 01 00:00:00 EDT 2018
Functional interplay between liver X receptor and AMP-activated protein kinase α inhibits atherosclerosis in apolipoprotein E-deficient mice - a new anti-atherogenic strategy.
British journal of pharmacology
Ma C,Zhang W,Yang X,Liu Y,Liu L,Feng K,Zhang X,Yang S,Sun L,Yu M,Yang J,Li X,Hu W,Miao RQ,Zhu Y,Li L,Han J,Chen Y,Duan Y
Tue May 01 00:00:00 EDT 2018
Functional interplay between liver X receptor and AMP-activated protein kinase α inhibits atherosclerosis in apolipoprotein E-deficient mice - a new anti-atherogenic strategy.
British journal of pharmacology
Ma C,Zhang W,Yang X,Liu Y,Liu L,Feng K,Zhang X,Yang S,Sun L,Yu M,Yang J,Li X,Hu W,Miao RQ,Zhu Y,Li L,Han J,Chen Y,Duan Y
Tue May 01 00:00:00 EDT 2018
Baicalein protects human vitiligo melanocytes from oxidative stress through activation of NF-E2-related factor2 (Nrf2) signaling pathway.
Free radical biology & medicine
Ma J,Li S,Zhu L,Guo S,Yi X,Cui T,He Y,Chang Y,Liu B,Li C,Jian Z
Sat Dec 01 00:00:00 EST 2018
Baicalein protects human vitiligo melanocytes from oxidative stress through activation of NF-E2-related factor2 (Nrf2) signaling pathway.
Free radical biology & medicine
Ma J,Li S,Zhu L,Guo S,Yi X,Cui T,He Y,Chang Y,Liu B,Li C,Jian Z
Sat Dec 01 00:00:00 EST 2018
Ubiquitination Regulates the Proteasomal Degradation and Nuclear Translocation of the Fat Mass and Obesity-Associated (FTO) Protein.
Journal of molecular biology
Zhu T,Yong XLH,Xia D,Widagdo J,Anggono V
Fri Feb 02 00:00:00 EST 2018
Ubiquitination Regulates the Proteasomal Degradation and Nuclear Translocation of the Fat Mass and Obesity-Associated (FTO) Protein.
Journal of molecular biology
Zhu T,Yong XLH,Xia D,Widagdo J,Anggono V
Fri Feb 02 00:00:00 EST 2018
Calycosin alleviates allergic contact dermatitis by repairing epithelial tight junctions via down-regulating HIF-1α.
Journal of cellular and molecular medicine
Jia Z,Wang X,Wang X,Wei P,Li L,Wu P,Hong M
Sat Sep 01 00:00:00 EDT 2018
Calycosin alleviates allergic contact dermatitis by repairing epithelial tight junctions via down-regulating HIF-1α.
Journal of cellular and molecular medicine
Jia Z,Wang X,Wang X,Wei P,Li L,Wu P,Hong M
Sat Sep 01 00:00:00 EDT 2018
Paracrine HGF/c-MET enhances the stem cell-like potential and glycolysis of pancreatic cancer cells via activation of YAP/HIF-1α.
Experimental cell research
Yan B,Jiang Z,Cheng L,Chen K,Zhou C,Sun L,Qian W,Li J,Cao J,Xu Q,Ma Q,Lei J
Mon Oct 01 00:00:00 EDT 2018
Paracrine HGF/c-MET enhances the stem cell-like potential and glycolysis of pancreatic cancer cells via activation of YAP/HIF-1α.
Experimental cell research
Yan B,Jiang Z,Cheng L,Chen K,Zhou C,Sun L,Qian W,Li J,Cao J,Xu Q,Ma Q,Lei J
Mon Oct 01 00:00:00 EDT 2018
SKP2 Promotes Hepatocellular Carcinoma Progression Through Nuclear AMPK-SKP2-CARM1 Signaling Transcriptionally Regulating Nutrient-Deprived Autophagy Induction.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Wei X,Li X,Yan W,Zhang X,Sun Y,Zhang F
Fri Aug 17 00:00:00 EDT 2018
SKP2 Promotes Hepatocellular Carcinoma Progression Through Nuclear AMPK-SKP2-CARM1 Signaling Transcriptionally Regulating Nutrient-Deprived Autophagy Induction.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Wei X,Li X,Yan W,Zhang X,Sun Y,Zhang F
Fri Aug 17 00:00:00 EDT 2018
Activation of hepatic Nogo-B receptor expression-A new anti-liver steatosis mechanism of statins.
Biochimica et biophysica acta. Molecular and cell biology of lipids
Zhang W,Yang X,Chen Y,Hu W,Liu L,Zhang X,Liu M,Sun L,Liu Y,Yu M,Li X,Li L,Zhu Y,Miao QR,Han J,Duan Y
Thu Feb 01 00:00:00 EST 2018
Role of c-Abl-GSK3β Signaling in MPP+-Induced Autophagy-Lysosomal Dysfunction.
Toxicological sciences : an official journal of the Society of Toxicology
Ren Y,Chen J,Wu X,Gui C,Mao K,Zou F,Li W
Sat Sep 01 00:00:00 EDT 2018
Role of c-Abl-GSK3β Signaling in MPP+-Induced Autophagy-Lysosomal Dysfunction.
Toxicological sciences : an official journal of the Society of Toxicology
Ren Y,Chen J,Wu X,Gui C,Mao K,Zou F,Li W
Sat Sep 01 00:00:00 EDT 2018
PADI2-Mediated Citrullination Promotes Prostate Cancer Progression.
Cancer research
Wang L,Song G,Zhang X,Feng T,Pan J,Chen W,Yang M,Bai X,Pang Y,Yu J,Han J,Han B
Wed Nov 01 00:00:00 EDT 2017
PADI2-Mediated Citrullination Promotes Prostate Cancer Progression.
Cancer research
Wang L,Song G,Zhang X,Feng T,Pan J,Chen W,Yang M,Bai X,Pang Y,Yu J,Han J,Han B
Wed Nov 01 00:00:00 EDT 2017
Nuclear retention of the lncRNA SNHG1 by doxorubicin attenuates hnRNPC-p53 protein interactions.
EMBO reports
Shen Y,Liu S,Fan J,Jin Y,Tian B,Zheng X,Fu H
Sat Apr 01 00:00:00 EDT 2017
Nuclear retention of the lncRNA SNHG1 by doxorubicin attenuates hnRNPC-p53 protein interactions.
EMBO reports
Shen Y,Liu S,Fan J,Jin Y,Tian B,Zheng X,Fu H
Sat Apr 01 00:00:00 EDT 2017
DExD/H-Box Helicase 36 Signaling via Myeloid Differentiation Primary Response Gene 88 Contributes to NF-κB Activation to Type 2 Porcine Reproductive and Respiratory Syndrome Virus Infection.
Frontiers in immunology
Jing H,Zhou Y,Fang L,Ding Z,Wang D,Ke W,Chen H,Xiao S
Wed Nov 20 00:00:00 EST 2019
DExD/H-Box Helicase 36 Signaling via Myeloid Differentiation Primary Response Gene 88 Contributes to NF-κB Activation to Type 2 Porcine Reproductive and Respiratory Syndrome Virus Infection.
Frontiers in immunology
Jing H,Zhou Y,Fang L,Ding Z,Wang D,Ke W,Chen H,Xiao S
Wed Nov 20 00:00:00 EST 2019
Cluster of Differentiation 36 Deficiency Aggravates Macrophage Infiltration and Hepatic Inflammation by Upregulating Monocyte Chemotactic Protein-1 Expression of Hepatocytes Through Histone Deacetylase 2-Dependent Pathway.
Antioxidants & redox signaling
Zhong S,Zhao L,Wang Y,Zhang C,Liu J,Wang P,Zhou W,Yang P,Varghese Z,Moorhead JF,Chen Y,Ruan XZ
Tue Aug 01 00:00:00 EDT 2017
Cluster of Differentiation 36 Deficiency Aggravates Macrophage Infiltration and Hepatic Inflammation by Upregulating Monocyte Chemotactic Protein-1 Expression of Hepatocytes Through Histone Deacetylase 2-Dependent Pathway.
Antioxidants & redox signaling
Zhong S,Zhao L,Wang Y,Zhang C,Liu J,Wang P,Zhou W,Yang P,Varghese Z,Moorhead JF,Chen Y,Ruan XZ
Tue Aug 01 00:00:00 EDT 2017
CRB3 downregulation confers breast cancer stem cell traits through TAZ/β-catenin.
Oncogenesis
Li P,Wang Y,Mao X,Jiang Y,Liu J,Li J,Wang J,Wang R,She J,Zhang J,Yang J,Liu Y,Liu P
Mon Apr 24 00:00:00 EDT 2017
CRB3 downregulation confers breast cancer stem cell traits through TAZ/β-catenin.
Oncogenesis
Li P,Wang Y,Mao X,Jiang Y,Liu J,Li J,Wang J,Wang R,She J,Zhang J,Yang J,Liu Y,Liu P
Mon Apr 24 00:00:00 EDT 2017
TRAIL facilitates cytokine expression and macrophage migration during hypoxia/reoxygenation via ER stress-dependent NF-κB pathway.
Molecular immunology
Jiang Y,Chen X,Fan M,Li H,Zhu W,Chen X,Cao C,Xu R,Wang Y,Ma Y
Wed Feb 01 00:00:00 EST 2017
TRAIL facilitates cytokine expression and macrophage migration during hypoxia/reoxygenation via ER stress-dependent NF-κB pathway.
Molecular immunology
Jiang Y,Chen X,Fan M,Li H,Zhu W,Chen X,Cao C,Xu R,Wang Y,Ma Y
Wed Feb 01 00:00:00 EST 2017
TRIM52: A nuclear TRIM protein that positively regulates the nuclear factor-kappa B signaling pathway.
Molecular immunology
Fan W,Liu T,Li X,Zhou Y,Wu M,Cui X,Chen H,Qian P
Wed Feb 01 00:00:00 EST 2017
TRIM52: A nuclear TRIM protein that positively regulates the nuclear factor-kappa B signaling pathway.
Molecular immunology
Fan W,Liu T,Li X,Zhou Y,Wu M,Cui X,Chen H,Qian P
Wed Feb 01 00:00:00 EST 2017
Highly expressed lncRNA CCND2-AS1 promotes glioma cell proliferation through Wnt/β-catenin signaling.
Biochemical and biophysical research communications
Zhang H,Wei DL,Wan L,Yan SF,Sun YH
Sun Jan 22 00:00:00 EST 2017
Highly expressed lncRNA CCND2-AS1 promotes glioma cell proliferation through Wnt/β-catenin signaling.
Biochemical and biophysical research communications
Zhang H,Wei DL,Wan L,Yan SF,Sun YH
Sun Jan 22 00:00:00 EST 2017
ERK1/2-mediated Cytoplasmic Accumulation of hnRNPK Antagonizes TRAIL-induced Apoptosis through Upregulation of XIAP in H1299 Cells.
Biomedical and environmental sciences : BES
Huang WS,Xu FM,Zeng QZ,Liu XH,Gao XJ,Liu LX
Sat Jul 01 00:00:00 EDT 2017
ERK1/2-mediated Cytoplasmic Accumulation of hnRNPK Antagonizes TRAIL-induced Apoptosis through Upregulation of XIAP in H1299 Cells.
Biomedical and environmental sciences : BES
Huang WS,Xu FM,Zeng QZ,Liu XH,Gao XJ,Liu LX
Sat Jul 01 00:00:00 EDT 2017
CRB3 regulates contact inhibition by activating the Hippo pathway in mammary epithelial cells.
Cell death & disease
Mao X,Li P,Wang Y,Liang Z,Liu J,Li J,Jiang Y,Bao G,Li L,Zhu B,Ren Y,Zhao X,Zhang J,Liu Y,Yang J,Liu P
Thu Jan 12 00:00:00 EST 2017
CRB3 regulates contact inhibition by activating the Hippo pathway in mammary epithelial cells.
Cell death & disease
Mao X,Li P,Wang Y,Liang Z,Liu J,Li J,Jiang Y,Bao G,Li L,Zhu B,Ren Y,Zhao X,Zhang J,Liu Y,Yang J,Liu P
Thu Jan 12 00:00:00 EST 2017
Regulation of hepatic lipogenesis by the zinc finger protein Zbtb20.
Nature communications
Liu G,Zhou L,Zhang H,Chen R,Zhang Y,Li L,Lu JY,Jiang H,Liu D,Qi S,Jiang YM,Yin K,Xie Z,Shi Y,Liu Y,Cao X,Chen YX,Zou D,Zhang WJ
Wed Mar 22 00:00:00 EDT 2017
Regulation of hepatic lipogenesis by the zinc finger protein Zbtb20.
Nature communications
Liu G,Zhou L,Zhang H,Chen R,Zhang Y,Li L,Lu JY,Jiang H,Liu D,Qi S,Jiang YM,Yin K,Xie Z,Shi Y,Liu Y,Cao X,Chen YX,Zou D,Zhang WJ
Wed Mar 22 00:00:00 EDT 2017
Dicer interacts with SIRT7 and regulates H3K18 deacetylation in response to DNA damaging agents.
Nucleic acids research
Zhang PY,Li G,Deng ZJ,Liu LY,Chen L,Tang JZ,Wang YQ,Cao ST,Fang YX,Wen F,Xu Y,Chen X,Shi KQ,Li WF,Xie C,Tang KF
Thu May 05 00:00:00 EDT 2016
Dicer interacts with SIRT7 and regulates H3K18 deacetylation in response to DNA damaging agents.
Nucleic acids research
Zhang PY,Li G,Deng ZJ,Liu LY,Chen L,Tang JZ,Wang YQ,Cao ST,Fang YX,Wen F,Xu Y,Chen X,Shi KQ,Li WF,Xie C,Tang KF
Thu May 05 00:00:00 EDT 2016
Biallelic SUN5 Mutations Cause Autosomal-Recessive Acephalic Spermatozoa Syndrome.
American journal of human genetics
Zhu F,Wang F,Yang X,Zhang J,Wu H,Zhang Z,Zhang Z,He X,Zhou P,Wei Z,Gecz J,Cao Y
Thu Oct 06 00:00:00 EDT 2016
Biallelic SUN5 Mutations Cause Autosomal-Recessive Acephalic Spermatozoa Syndrome.
American journal of human genetics
Zhu F,Wang F,Yang X,Zhang J,Wu H,Zhang Z,Zhang Z,He X,Zhou P,Wei Z,Gecz J,Cao Y
Thu Oct 06 00:00:00 EDT 2016
Novel function of PITH domain-containing 1 as an activator of internal ribosomal entry site to enhance RUNX1 expression and promote megakaryocyte differentiation.
Cellular and molecular life sciences : CMLS
Lu B,Sun X,Chen Y,Jin Q,Liang Q,Liu S,Li Y,Zhou Y,Li W,Huang Z
Sun Feb 01 00:00:00 EST 2015
Novel function of PITH domain-containing 1 as an activator of internal ribosomal entry site to enhance RUNX1 expression and promote megakaryocyte differentiation.
Cellular and molecular life sciences : CMLS
Lu B,Sun X,Chen Y,Jin Q,Liang Q,Liu S,Li Y,Zhou Y,Li W,Huang Z
Sun Feb 01 00:00:00 EST 2015
Set7 facilitates hepatitis C virus replication via enzymatic activity-dependent attenuation of the IFN-related pathway.
Journal of immunology (Baltimore, Md. : 1950)
Han T,Wan Y,Wang J,Zhao P,Yuan Y,Wang L,She Y,Broering R,Lu M,Ye L,Zhu Y
Sun Mar 15 00:00:00 EDT 2015
Set7 facilitates hepatitis C virus replication via enzymatic activity-dependent attenuation of the IFN-related pathway.
Journal of immunology (Baltimore, Md. : 1950)
Han T,Wan Y,Wang J,Zhao P,Yuan Y,Wang L,She Y,Broering R,Lu M,Ye L,Zhu Y
Sun Mar 15 00:00:00 EDT 2015
miR-206 modulates lipopolysaccharide-mediated inflammatory cytokine production in human astrocytes.
Cellular signalling
Duan X,Zohaib A,Li Y,Zhu B,Ye J,Wan S,Xu Q,Song Y,Chen H,Cao S
Thu Jan 01 00:00:00 EST 2015
miR-206 modulates lipopolysaccharide-mediated inflammatory cytokine production in human astrocytes.
Cellular signalling
Duan X,Zohaib A,Li Y,Zhu B,Ye J,Wan S,Xu Q,Song Y,Chen H,Cao S
Thu Jan 01 00:00:00 EST 2015
Novel function of the chromosome 7 open reading frame 41 gene to promote leukemic megakaryocyte differentiation by modulating TPA-induced signaling.
Blood cancer journal
Sun X,Lu B,Hu B,Xiao W,Li W,Huang Z
Fri Mar 28 00:00:00 EDT 2014
Novel function of the chromosome 7 open reading frame 41 gene to promote leukemic megakaryocyte differentiation by modulating TPA-induced signaling.
Blood cancer journal
Sun X,Lu B,Hu B,Xiao W,Li W,Huang Z
Fri Mar 28 00:00:00 EDT 2014
UDP-glucose dehydrogenase modulates proteoglycan synthesis in articular chondrocytes: its possible involvement and regulation in osteoarthritis.
Arthritis research & therapy
Wen Y,Li J,Wang L,Tie K,Magdalou J,Chen L,Wang H
Wed Dec 03 00:00:00 EST 2014
UDP-glucose dehydrogenase modulates proteoglycan synthesis in articular chondrocytes: its possible involvement and regulation in osteoarthritis.
Arthritis research & therapy
Wen Y,Li J,Wang L,Tie K,Magdalou J,Chen L,Wang H
Wed Dec 03 00:00:00 EST 2014
FHL2-driven molecular network mediated Septin2 knockdown inducing apoptosis in mesangial cell.
Proteomics
Lu Y,Cai G,Cui S,Geng W,Chen D,Wen J,Zhang Y,Zhang F,Xie Y,Fu B,Chen X
Sat Nov 01 00:00:00 EDT 2014
FHL2-driven molecular network mediated Septin2 knockdown inducing apoptosis in mesangial cell.
Proteomics
Lu Y,Cai G,Cui S,Geng W,Chen D,Wen J,Zhang Y,Zhang F,Xie Y,Fu B,Chen X
Sat Nov 01 00:00:00 EDT 2014
Protective effect of astaxanthin on liver fibrosis through modulation of TGF-β1 expression and autophagy.
Mediators of inflammation
Shen M,Chen K,Lu J,Cheng P,Xu L,Dai W,Wang F,He L,Zhang Y,Chengfen W,Li J,Yang J,Zhu R,Zhang H,Zheng Y,Zhou Y,Guo C
Thu Dec 18 00:00:00 EST 2014
Protective effect of astaxanthin on liver fibrosis through modulation of TGF-β1 expression and autophagy.
Mediators of inflammation
Shen M,Chen K,Lu J,Cheng P,Xu L,Dai W,Wang F,He L,Zhang Y,Chengfen W,Li J,Yang J,Zhu R,Zhang H,Zheng Y,Zhou Y,Guo C
Thu Dec 18 00:00:00 EST 2014
Proteomic analysis of primary duck hepatocytes infected with duck hepatitis B virus.
Proteome science
Zhao Y,Ben H,Qu S,Zhou X,Yan L,Xu B,Zhou S,Lou Q,Ye R,Zhou T,Yang P,Qu D
Mon Jun 07 00:00:00 EDT 2010
Proteomic analysis of primary duck hepatocytes infected with duck hepatitis B virus.
Proteome science
Zhao Y,Ben H,Qu S,Zhou X,Yan L,Xu B,Zhou S,Lou Q,Ye R,Zhou T,Yang P,Qu D
Mon Jun 07 00:00:00 EDT 2010
Proteomic analysis of protein complexes in human SH-SY5Y neuroblastoma cells by using blue-native gel electrophoresis: an increase in lamin A/C associated with heat shock protein 90 in response to 6-hydroxydopamine-induced oxidative stress.
Experimental gerontology
Nakamura M,Morisawa H,Imajoh-Ohmi S,Takamura C,Fukuda H,Toda T
Fri Nov 05 00:00:00 EDT 2010
Proteomic analysis of protein complexes in human SH-SY5Y neuroblastoma cells by using blue-native gel electrophoresis: an increase in lamin A/C associated with heat shock protein 90 in response to 6-hydroxydopamine-induced oxidative stress.
Experimental gerontology
Nakamura M,Morisawa H,Imajoh-Ohmi S,Takamura C,Fukuda H,Toda T
Fri Nov 05 00:00:00 EDT 2010