Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Samples: HepG2, Jurkat, HT-29, U-251MG, A-549, Mouse spleen, Rat testis
Immunogen sequence: RVTVSRASSS RSVRTTRGKR KRVDVEESEA SSSVSISHSA SATGNVCIEE IDVDGKFIRL KNTSEQDQPM GGWEMIRKIG DTSVSYKYTS RYVLKAGQTV TIWAANAGVT ASPPTDLIWK NQNSWGTGED VKVILKNSQG EEVAQRSTVF KTTIPEEEEE EEEAAGVVVE EELFHQQGTP RASNRSCAIM
Laminins, a family of extracellular matrix glycoproteins, are the major noncollagenous constituent of basement membranes. They have been implicated in a wide variety of biological processes including cell adhesion, differentiation, migration, signaling, neurite outgrowth and metastasis. Laminins are composed of 3 non identical chains: laminin alpha, beta and gamma (formerly A, B1, and B2, respectively) and they form a cruciform structure consisting of 3 short arms, each formed by a different chain, and a long arm composed of all 3 chains. Defects in the LMNB1 gene can cause Leukodystrophy, demyelinating, autosomal dominant, adult-onset (ADLD).
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Lamin-B1; MGC111419
Gene Aliases: ADLD; LMN; LMN2; LMNB; LMNB1
UniProt ID: (Human) P20700, (Mouse) P14733, (Rat) P70615
Entrez Gene ID: (Human) 4001, (Mouse) 16906, (Rat) 116685
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support