Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Laminin is a non-collagenous protein of basement membranes. By rotary shadowing it has a cross-like shape, consisting of one long arm, (400 kD) and three short arms (200 kD). Antibodies to laminin localize exclusively in basement membranes. Laminin is found throughout the basement membrane, but the reaction with the antibodies to laminin is more intense in the lamina lucida, the part of the basement membrane adjacent to the cell membrane. Laminin binds to type IV collagen, a component exclusively localized in the basement membrane. Laminin has been shown to play a role in cell adhesion and attachment in vivo and in vitro.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dystrophia muscularis; laminin alpha-2; laminin M; Laminin M chain; Laminin subunit alpha-2; laminin, alpha 2; Laminin-12 subunit alpha; Laminin-2 subunit alpha; Laminin-4 subunit alpha; Merosin heavy chain
Gene Aliases: 5830440B04; dy; LAMA2; LAMM; mer; merosin; mKIAA4087
UniProt ID: (Human) P24043, (Mouse) Q60675
Entrez Gene ID: (Human) 3908, (Rat) 309368, (Mouse) 16773
Molecular Function:
extracellular matrix protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support