Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive test controls include: SW480, HepG2, HT-1080, SKOV3, Mouse heart. The target is usually found in the following locations: Secreted, basement membrane, extracellular matrix, extracellular space.
Immunogen sequence: EAKLRADEAK QSAEDILLKT NATKEKMDKS NEELRNLIKQ IRNFLTQDSA DLDSIEAVAN EVLKMEMPST PQQLQNLTED IRERVESLSQ VEVILQHSAA DIARAEMLLE EAKRASKSAT DVKVTADMVK EALEEAEKAQ VAAEKAIKQA DEDIQGTQNL LTSIESETAA SEETLFNASQ RISELERNVE ELKRKAAQNS GEAEYIEKVV YTVKQSAEDV KKTLDGELDE KYKKVENLIA KKTEESADAR RKAEMLQNEA KTLLAQANSK LQLLKDLERK YEDNQRYLED KAQELARLEG EVRSLLKDIS QKVAVYSTCL
Laminins are large heterotrimeric, noncollagenous glycoproteins composed of alpha, beta, and gamma chains. They are ubiquitously present in basement membrane (BM) along with entactin/nidogen (EN), collagen type IV (CIV), and large heparan sulfate proteoglycan (HSPG), which interact specifically with each other to form a continuous and regular BM. Alterations of BM integrity, from local discontinuities up to complete loss, are described in many types of human and animal epithelial neoplasms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Laminin B1 chain; laminin B1 subunit 1; laminin beta 1; Laminin subunit beta-1; laminin, beta 1; Laminin-1 subunit beta; Laminin-10 subunit beta; Laminin-12 subunit beta; Laminin-2 subunit beta; Laminin-6 subunit beta; Laminin-8 subunit beta; MGC142015
Gene Aliases: C77966; C80098; C81607; CLM; D130003D08Rik; Lamb-1; LAMB1; Lamb1-1; LIS5
UniProt ID: (Human) P07942, (Mouse) P02469
Entrez Gene ID: (Human) 3912, (Mouse) 16777, (Rat) 298941
Molecular Function:
extracellular matrix protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support