Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 2
Clone M046 detects native laminin isoforms that include both laminin beta2 and gamma1 subunits. In native immunoblots, the antibody detects laminin 521, laminin 221, and laminin 121, but does not detect lamin 332 and laminin 411. The antibody weakly detects laminin 511. This antibody can be used for immunocytochemical detection of laminin beta2/gamma1 subunits, and can also be used for detection of native laminin isoforms in immunoblots and ELISA.
Laminins are large heterotrimeric, noncollagenous glycoproteins composed of alpha, beta, and gamma chains. They are ubiquitously present in basement membrane (BM) along with entactin/nidogen (EN), collagen type IV (CIV), and large heparan sulfate proteoglycan (HSPG), which interact specifically with each other to form a continuous and regular BM. Alterations of BM integrity, from local discontinuities up to complete loss, are described in many types of human and animal epithelial neoplasms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AA408497; Lama; Laminin B1s chain; Laminin B2 chain; laminin S; Laminin subunit beta-2; Laminin subunit gamma-1; laminin, beta 2 (laminin S); laminin, gamma 1 (formerly LAMB2); Laminin-1 subunit gamma; Laminin-10 subunit gamma; Laminin-11 subunit beta; Laminin-11 subunit gamma; Laminin-14 subunit beta; Laminin-15 subunit beta; Laminin-2 subunit gamma; Laminin-3 subunit beta; Laminin-3 subunit gamma; Laminin-4 subunit beta; Laminin-4 subunit gamma; Laminin-6 subunit gamma; Laminin-7 subunit beta; Laminin-7 subunit gamma; Laminin-8 subunit gamma; Laminin-9 subunit beta; Laminin-9 subunit gamma; S-LAM beta; S-LAM gamma; S-laminin subunit beta; S-laminin subunit gamma
Gene Aliases: LAMB2; LAMC1; LAMS; NPHS5
UniProt ID: (Human) P55268, (Human) P11047
Entrez Gene ID: (Human) 3913, (Human) 3915
Molecular Function:
extracellular matrix protein
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support