Invitrogen
FIGURE: 1 / 2
Store as concentrated solution.
Centrifuge briefly prior to opening vial.
Lassa fever virus, a member of the family Arenaviridae, is a highly endemic category A pathogen that causes 300,000-500,000 infections per year in Western Africa. The arenaviral nucleoprotein NP has been implicated in suppression of the host innate immune system.
Protein Aliases: Lassa mammarenavirus NP; Lassa mammarenavirus Nucleoprotein; Lassa virus NP; Lassa virus Nucleoprotein; LASV NP; LASV Nucleoprotein