Search Thermo Fisher Scientific
Exalpha
Positive control: Purified luciferase protein.
Detects luciferase protein by Western blot in C. elegans and Drosophilia melanogaster tissues, human fibroblast, mouse macrophage, kidney, liver and cortex as well as NIH3T3, Jurkat and BHR21 cell lines. Detects luciferase with little to no background signal.
Luciferase produces green light with the wavelength of 562 nm. Luciferase catalyzes the reaction: D-firefly luciferin + O2 + ATP = firefly oxyluciferin + CO2 + AMP + diphosphate + light.
Protein Aliases: Firefly Luciferase; Genji firefly; Green Luciferase; Japanese firefly; luciferin 4-monooxygenase; Photinus pyralis; photinus pyralis luciferase