Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Ly108 is the mouse homologue of human SLAMF6 (signaling lymphocyte activation molecule family member 6). Mouse Ly108 is expressed on NK, B cells, and T cells. Activation of Ly108 receptor enhances NK cell activity and IFN-gamma expression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activating NK receptor; CD352; Lymphocyte antigen 108; natural killer-, T- and B-cell antigen; NK-T-B-antigen; NTB-A; NTBA receptor; SLAM family member 6
Gene Aliases: CD352; KAL1; KAL1b; KALI; KALIb; Ly108; NTB-A; NTBA; RGD1561848; SF2000; SLAMF6; UNQ6123/PRO20080
UniProt ID: (Human) Q96DU3, (Mouse) Q9ET39
Entrez Gene ID: (Human) 114836, (Mouse) 30925, (Rat) 498287
Molecular Function: immunoglobulin receptor superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support