Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Description: The AT8 monoclonal antibody reacts with the mouse Ly-49G receptor from 129, FVB, BALB/c, C3H, CBA, SJL and A/J, but not C57Bl/6 mice. Ly49G, a member of the Ly49 family is expressed by NK cell subset.
Applications Reported: The AT-8 (AT8) antibody has been reported for use in flow cytometric analysis.
Applications Tested: The AT-8 (AT8) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.
Filtration: 0.2 µm post-manufacturing filtered.
Mouse Ly49 receptors exhibit allelic specificity for MHC class I Ia molecules and are thought to serve to prevent natural killer (NK) cells from attacking normal cells, while allowing them to attack infected or transformed cells in which class I molecules have been down-regulated. These inhibitory receptors are also expressed on subpopulations of mouse CD8+ T cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.
Protein Aliases: Killer cell lectin-like receptor 7; Ly-49g; Lymphocyte antigen 49g; T-cell surface glycoprotein Ly-49G
Gene Aliases: Klra7; LGL-1; Ly-49g; Ly-49G.1; Ly-49G.2; Ly-49G.3; Ly49-g; Ly49g; Ly49G.4; Ly49g1; Ly49g2; Ly49g4
UniProt ID: (Mouse) Q60654
Entrez Gene ID: (Mouse) 16638
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support