Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: CELARTLKR LGMDGYRGIS LANWMCLAKW ESGYNTRATN YNAGDRSTDY GIFQINSRYW CNDGKTPGAV NACHLSCSAL LQDNIADAVA CAKRVVRDPQ GIRAWVAWRN RCQNRDVRQY VQGCGV (24-148 aa encoded by BC004147)
This gene encodes human lysozyme, whose natural substrate is the bacterial cell wall peptidoglycan (cleaving the beta[1-4]glycosidic linkages between N-acetylmuramic acid and N-acetylglucosamine). Lysozyme is one of the anti-microbial agents found in human milk, and is also present in spleen, lung, kidney, white blood cells, plasma, saliva, and tears. Missense mutations in LYZ have been identified in heritable renal amyloidosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1,4-beta-N-acetylmuramidase C; c-type lysozyme; lysozyme (renal amyloidosis); Lysozyme C; Lysozyme C type M; Lysozyme C type P; Lysozyme C-1; Lysozyme C-2; lysozyme F1; P lysozyme structural; renal amyloidosis
Gene Aliases: AI326280; Lys; Lysm; Lysz; LYZ; Lyz1; Lyz2; LYZF1; LYZS; LZM; Lzm-s1; Lzp; Lzp-s
UniProt ID: (Human) P61626, (Pig) P12068, (Mouse) P17897, (Mouse) P08905
Entrez Gene ID: (Human) 4069, (Pig) 100157211, (Mouse) 17110, (Mouse) 17105, (Rat) 25211
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support