Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
SNARE that acts to regulate protein transport between late endosomes and the trans-Golgi network. The SNARE complex containing STX6, STX12, VAMP4 and VTI1A mediates vesicle fusion (in vitro).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Actin cross-linking family 7; actin-crosslinking protein 7; FLJ45612; FLJ46776; macrophin 1; Microtubule-actin cross-linking factor 1; trabeculin alpha
Gene Aliases: ABP620; Acf7; Aclp7; MACF; Macf1; mKIAA0465; R74989
UniProt ID: (Rat) D3ZHV2
Entrez Gene ID: (Mouse) 11426, (Rat) 362587
Molecular Function:
intermediate filament binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support