Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MAX dimerization protein belongs to a subfamily of MAX-interacting proteins. This protein competes with MYC for binding to MAX to form a sequence-specific DNA-binding complex, acts as a transcriptional repressor (while MYC appears to function as an activator) and is a candidate tumor suppressor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hMAD1; HsMAD1; MAD1 mitotic arrest deficient-like 1; MAD1-like protein 1; mitotic arrest deficient 1-like 1; Mitotic arrest deficient 1-like protein 1; Mitotic checkpoint MAD1 protein homolog; Mitotic spindle assembly checkpoint protein MAD1; mitotic-arrest deficient 1, yeast, homolog-like 1; MXD1; Tax-binding protein 181; tumor protein p53 inducible protein 9
Gene Aliases: AI173502; AW550425; MAD1; MAD1L1; PIG9; TP53I9; TXBP181
UniProt ID: (Human) Q9Y6D9, (Mouse) Q9WTX8
Entrez Gene ID: (Human) 8379, (Rat) 680006, (Mouse) 17120
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support