Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MAAPEAEVLS SAAVPDLEWY EKSEETHASQ IELLETSSTQ EPLNASEAFC PRDCMVPVVF PGPVSQEGCC QFTCELLKHI MYQRQQLPLP YEQLKHFYRK PSPQAEEMLK KKPRATTEVS SRKCQQALAE LESVLSHLED FFARTLVPRV LILLGGNALS PKEFYELDLS LLAPYSVDQS LSTAACLRRL FRAIFMADAF SELQAPPLMG TVVMAQGHRN CGEDWFRPKL NYRVPSRGHK LTVTLSCGRP SIRTTAWEDY IWFQAPVTFK GFRE
The protein encoded by this gene was identified as a binding protein of the MAD2 mitotic arrest deficient-like 1 (MAD2/MAD2L1). MAD2 is a key component of the spindle checkpoint that delays the onset of anaphase until all the kinetochores are attached to the spindle. This protein may interact with the spindle checkpoint and coordinate cell cycle events in late mitosis. Alternatively spliced transcript variants encoding distinct isoforms have been observed.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Caught by MAD2 protein; MAD2L1-binding protein; p31(comet); RP1-261G23.6
Gene Aliases: CMT2; KIAA0110; MAD2L1BP
UniProt ID: (Human) Q15013
Entrez Gene ID: (Human) 9587
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support