Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAVQESAAQ LSMTLKVQEY PTLKVPYETL NKRFRAAQKN IDRETSHVTM VVAELEKTLS GCPAVDSVVS LLDGVVEKLS VLKRKAVESI QAEDESAKLC KRRIEHLKEH SSDQPAAASV WKRKRMDRMM VEHLLRCGYY NTAVKLARQS GIEDLVNIEM FLTAKEVEES LERRETATCL AWCHDNKSRL RKMKSCLEFS LRIQEFIELI RQNKRLDAVR HARKHFSQAE GSQLDEVRQA MGMLAFPPDT HISPYKDLLD PARWRMLIQQ FRYDNYRLHQ LGNNSVFTLT LQAGLSAIKT PQCYKEDGSS KSPDCPVCSR SLNKLAQPLP MAHCANSRLV CKISGDVMNE NNPPMMLPNG YVYGYNSLLS IRQDDKVVCP RTKEVFHFSQ AEKVYIM (1-396 aa encoded by BC001225)
This gene product mediates the attachment of erythroblasts to macrophages. This attachment promotes terminal maturation and enucleation of erythroblasts, presumably by suppressing apoptosis. This protein is an integral membrane protein with the N-terminus on the extracellular side and the C-terminus on the cytoplasmic side of the cell. Two immunologically related isoforms of erythroblast macrophage protein with apparent molecular weights of 33 kD and 36 kD were detected in macrophage membranes, this gene encodes the larger isoform. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene, but the biological validity of some variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell proliferation-inducing gene 5 protein; E3 ubiquitin-protein transferase MAEA; Erythroblast macrophage protein; GID complex subunit 9, FYV10 homolog; HLC-10; Human lung cancer oncogene 10 protein; lung cancer-related protein 10; Macrophage erythroblast attacher; P44EMLP
Gene Aliases: EMLP; EMP; GID9; HLC-10; HLC10; MAEA; PIG5
UniProt ID: (Human) Q7L5Y9
Entrez Gene ID: (Human) 10296
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support