Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPVGGLLPLF SSPAGGVLGG GLGGGGGRKG SGPAALRLTE KFVLLLVFSA FITLCFGAIF FLPDSSKLLS GVLFHSSPAL QPAADHKPGP GARAEDAAEG RARRREEGAP GDPEAALEDN LARIRENHER ALREAKETLQ KLPEEIQRDI LLEKKKVAQD QLRDKAPFRG LPPVDFVPPI GVESREPADA AIREKRAKIK EEIEARKGQA NCPGSSSSCE VEIQMMKHAW NNYKGYAWGL NELKPISKGG HSSSLFGNIK GATIVDALDT LFIMEMKHEF EEAKSWVEEN LDFNVNAEIS VFEVNIRFVG GLLSAYYLSG EEIFRKKAVE LGVKLLPAFH TPSGIPWALL NMKRWMCNIP RAMEATI
This gene encodes a class I mammalian Golgi 1,2-mannosidase which is a type II transmembrane protein. This protein catalyzes the hydrolysis of three terminal mannose residues from peptide-bound Man(9)-GlcNAc(2) oligosaccharides and belongs to family 47 of glycosyl hydrolases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-1,2-mannosidase IA; Man(9)-alpha-mannosidase; Man9-mannosidase; Mannosidase alpha class 1A member 1; mannosidase, alpha, class 1A, member 1; Mannosyl-oligosaccharide 1,2-alpha-mannosidase IA; Processing alpha-1,2-mannosidase IA
Gene Aliases: HUMM3; HUMM9; MAN1A1; MAN9
UniProt ID: (Human) P33908
Entrez Gene ID: (Human) 4121
Molecular Function:
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support