Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Western blot analysis in human brain using this antibody detected two main bands between 100-130 kDa corresponding to heavy and light chains of MAP1S.
Immunogen sequence: APTSEAGLS LPLRGPRARR SASPHDVDLC LVSPCEFEHR KAVPMAPAPA SPGSSNDSSA RSQERAGGLG AEETPPTSVS ESLPTLSDSD PVPLAPGAAD SDEDTEGFGV PRHDPLPDPL KVPPPLPDPS SICMVDPEML PPKTARQTEN VSRTRKPLAR PNSRAAAPKA TPVAAAKTKG LAGGDRASRP LSARSEPSEK GGRAPLSRKS STPKTATRGP SGSASSRPGV SATPPKSPVY LDLAYLPSGS SAHLVDEEFF QRVRALCYVI SGQDQRKEEG MRAVLDALLA SKQHWDRDLQ VTLIPTFDSV AMHTWYAETH ARHQALGITV LGSNSMVSMQ DDAFPACKVE F (457-806 aa encoded by BC008806)
Microtubule-associated protein that mediates aggregation of mitochondria resulting in cell death and genomic destruction (MAGD). Plays a role in anchoring the microtubule organizing center to the centrosomes. Binds to DNA. Plays a role in apoptosis. Involved in the formation of microtubule bundles.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BPY2 interacting protein 1; BPY2-interacting protein 1; MAP-1S; Microtubule-associated protein 1S; Microtubule-associated protein 8; Variable charge Y chromosome 2-interacting protein 1; VCY2 interacting protein 1; VCY2-interacting protein 1
Gene Aliases: BPY2IP1; C19orf5; MAP1S; MAP8; VCY2IP-1; VCY2IP1
UniProt ID: (Human) Q66K74
Entrez Gene ID: (Human) 55201
Molecular Function:
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support