Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: AASRPSPSGG DSVLPKSISS AHDTRGSSVP ENDRLASIAA ELQFRSLSRH SSPTEERDEP AYPRGDSSGS TRRSWELRTL ISQSKDTASK LGPIEAIQKS
This gene encodes a TBP- (TATA box-binding protein) associated phosphoprotein that represses both basal and activated levels of transcription. The encoded protein is phosphorylated in vivo and this phosphorylation affects its interaction with TBP. This protein contains a histone fold motif at the amino terminus, a TBP-binding domain, and a glutamine- and alanine-rich region. The binding of DR1 repressor complexes to TBP-promoter complexes may establish a mechanism in which an altered DNA conformation, together with the formation of higher order complexes, inhibits the assembly of the preinitiation complex and controls the rate of RNA polymerase II transcription.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dJ473J16.1 (mitogen-activated protein kinase kinase kinase 4); JNKK1; MAP three kinase 1; MAP/ERK kinase kinase 4; MAP2K4; MAPK/ERK kinase kinase 4; MEK kinase 4; Mitogen-activated protein kinase kinase kinase 4; predicted protein of HQ0412; PRKMK4; SERK1; SSK2/SSK22 MAP kinase kinase kinase, yeast, homolog of
Gene Aliases: KIAA0213; MAP3K4; MAPKKK4; MEKK 4; MEKK4; MTK1; PRO0412
UniProt ID: (Human) Q9Y6R4
Entrez Gene ID: (Human) 4216
Molecular Function: non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support